Skip to Content
Merck
All Photos(1)

Documents

46624

Supelco

Phenothiazine

VETRANAL®, analytical standard

Synonym(s):

10H-Phenothiazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9NS
CAS Number:
Molecular Weight:
199.27
Beilstein:
143237
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

Assay

≥98%

form

powder

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

371 °C (lit.)

mp

182-187 °C (lit.)

application(s)

cleaning products
clinical
cosmetics
food and beverages
forensics and toxicology
personal care
pharmaceutical (small molecule)

format

neat

SMILES string

N1c2ccccc2Sc3ccccc13

InChI

1S/C12H9NS/c1-3-7-11-9(5-1)13-10-6-2-4-8-12(10)14-11/h1-8,13H

InChI key

WJFKNYWRSNBZNX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Blood

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alina N Sekretaryova et al.
Analytical chemistry, 84(3), 1220-1223 (2011-12-31)
Wiring glucose oxidase in the membrane with an immobilized mediator is possible due to the diffusion ability of the latter, if the enzyme containing membrane is formed according to the proposed protocol, including exposing proteins to water-organic mixtures with the
Sh Y Mamedov et al.
Bulletin of experimental biology and medicine, 150(5), 576-578 (2012-01-12)
We studied the effects of natural (α-tocopherol and SOD) and synthetic (potassium phenosane and hydroxypyridine) antioxidants on the recovery of disordered parameters of the electroretinogram under conditions of high-intensity light exposure. Potassium phenosane and tocopherol acetate more effectively normalized parameters
Daniella Takács et al.
Bioorganic & medicinal chemistry, 20(14), 4258-4270 (2012-06-29)
N-dienylphenothiazines synthesized from tetrazolo[1,5-a]pyridinium salts by treatment with phenothiazine were subjected to catalytic hydrogenation to yield N-butylphenothiazines, whereas transformation of these dienes with borane dimethyl sulfide (BH(3) × Me(2)S) resulted in selective hydroboration of one double bond and full reduction
V P Volkov
Klinicheskaia meditsina, 89(4), 27-30 (2011-09-22)
Ten most common and clinically significant pathological changes of ECG in schizophrenic patients with and without phenothiazine-induced cardiomyopathy (PCMP) at different stages of neuroleptic therapy are considered. A total of 46 men and 10 women were available for observation. The
Caiwen Ou et al.
Chemical communications (Cambridge, England), 49(18), 1853-1855 (2013-01-31)
We report a 'super-gelator' of a short peptide capped with phenothiazine acetic acid.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service