Skip to Content
Merck

46384

Florisil® Inorganic Adsorbent

30-60 mesh

Synonym(s):

Florisil®, Activated magnesium silicate, Magnesium silicate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
MgO3Si
CAS Number:
Molecular Weight:
100.39
NACRES:
SB.52
PubChem Substance ID:
UNSPSC Code:
23201100
EC Number:
215-681-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Florisil® Adsorbent for Chromatography, 30-60 mesh

grade

standard

Quality Segment

form

granular

technique(s)

LPLC: suitable

matrix

Magnesium silicate hydrate

matrix active group

magnesium silicate hydrate

particle size

250-595 μm, 30-60 mesh

pore size

60 Å mean pore size

separation technique

hydrophilic interaction (HILIC)

SMILES string

[Mg++].[O-][Si]([O-])=O

InChI

1S/Mg.O3Si/c;1-4(2)3/q+2;-2

InChI key

FKHIFSZMMVMEQY-UHFFFAOYSA-N

General description

Florisil, adsorbent for the separation of lipids, is a commercially-prepared magnesia silica gel. It has a coarse mesh size which helps in rapid flow rate. Florisil is effective in promoting [1,3]-sigmatropic shift of myceophenolic acid related allyl phenyl ethers. Florisil cartridges are used in atmospheric sampling to assess possible interference from air contaminants.

Application

Florisil was used:
  • as column packing material in column chromatography
  • as an adsorbent in Gas chromatography with electron capture detector (GC/ECD), capillary column chromatography, adsorption column chromatography
  • in clean-up procedures in gas chromatography-tandem mass spectrometry and GC-FID

Legal Information

Florisil is a registered trademark of U.S. Silica Company


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library