Skip to Content
Merck
All Photos(1)

Documents

46293

Supelco

Fenamiphos-sulfoxide

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H22NO4PS
CAS Number:
Molecular Weight:
319.36
Beilstein:
2390888
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCOP(=O)(NC(C)C)Oc1ccc(c(C)c1)S(C)=O

InChI

1S/C13H22NO4PS/c1-6-17-19(15,14-10(2)3)18-12-7-8-13(20(5)16)11(4)9-12/h7-10H,6H2,1-5H3,(H,14,15)

InChI key

LUQMWGMGWJEGAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Fenamiphos-sulfoxide is a metabolite of the pesticide fenamiphos, commonly found as a ground water contaminant.

Application

Fenamiphos-sulfoxide may be used as an analytical reference standard for the quantification of the analyte in environmental samples using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of Organophosphorus Pesticides in Water by Solid-Phase Extraction Followed by Liquid Chromatography/High-Flow Pneumatically Assisted Electrospray Mass Spectrometry
Molina.C, et al.
Analytical Chemistry, 66(24), 4444-4449 (1994)
Determination of National Survey of Pesticides analytes in groundwater by liquid chromatography with postcolumn reaction detection
Miles JC
Analytical Chemistry, 592(1-2), 283-290 (1992)
Simultaneous enantioselective determination of fenamiphos and its two metabolites in soil sample by CE
Lecoeur-Lorin M, et al.
Electrophoresis, 30(16), 283-290 (2009)
Effects of laboratory lighting on the stability of analytical grade pesticides.
K L Crepeau et al.
Bulletin of environmental contamination and toxicology, 45(3), 446-451 (1990-09-01)
Tanya P Cáceres et al.
Current microbiology, 57(6), 643-646 (2008-10-17)
The degradation of an organophosphorus pesticide, fenamiphos, by different species of five green algae and five cyanobacteria was studied. All the species tested were able to transform fenamiphos to its primary oxidation product, fenamiphos sulfoxide (FSO), while the majority of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service