Skip to Content
Merck

46037

Cyphenothrin

PESTANAL®, analytical standard, sum of isomers

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C24H25NO3
CAS Number:
Molecular Weight:
375.46
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
254-484-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

mixture of cis and trans isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(C)=C\C1C(C(=O)OC(C#N)c2cccc(Oc3ccccc3)c2)C1(C)C

InChI

1S/C24H25NO3/c1-16(2)13-20-22(24(20,3)4)23(26)28-21(15-25)17-9-8-12-19(14-17)27-18-10-6-5-7-11-18/h5-14,20-22H,1-4H3

InChI key

FJDPATXIBIBRIM-UHFFFAOYSA-N

General description

Cyphenothrin is classified under the synthetic pyrethroid category of pesticides.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Cyphenothrin may be used as a reference standard for the identification of cyphenothrin in:
  • Animal fat samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) mode of detection.
  • Indoor air by passive sampling using Tenax® TA tubes followed by thermal desorption gas chromatography-mass spectrometry (TD-GC-MS).
  • Urine samples by GC-MS with electron impact (EI) ionization.
  • Fruit matrices by HPLC with ultraviolet (UV) detection.
Cyphenothrin may be used as a reference standard for the identification of cyphenothrin in:
  • Animal fat samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionization (ESI) and multiple reaction monitoring (MRM) mode of detection.
  • Indoor air by passive sampling using Tenax Tenax® tubes followed by thermal desorption gas chromatography-mass spectrometry (TD-GC-MS).
  • Urine samples by GC-MS with electron impact (EI) ionization.
  • Fruit matrices by HPLC with ultraviolet (UV) detection.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Tenax is a registered trademark of Buchem B.V.


Still not finding the right product?

Explore all of our products under Cyphenothrin


pictograms

Exclamation markEnvironment

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library