Skip to Content
Merck

46013

Tribenuron-methyl

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H17N5O6S
CAS Number:
Molecular Weight:
395.39
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
401-190-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COC(=O)c1ccccc1S(=O)(=O)NC(=O)N(C)c2nc(C)nc(OC)n2

InChI

1S/C15H17N5O6S/c1-9-16-13(18-14(17-9)26-4)20(2)15(22)19-27(23,24)11-8-6-5-7-10(11)12(21)25-3/h5-8H,1-4H3,(H,19,22)

InChI key

VLCQZHSMCYCDJL-UHFFFAOYSA-N

General description

Tribenuron-methyl is a sulfonyl urea herbicide derivative employed for protection of crops such as corn, citrus, potatoes, tomatoes, rice and vines via inhibiting the growth of many grasses and broad-leaf weed species.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support. Tribenuron-methyl may be used as an analytical reference standard for the quantification of the analyte in environmental samples using different chromatography techniques.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Moisture sensitive, handle under argon.


Still not finding the right product?

Explore all of our products under Tribenuron-methyl


signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library