Skip to Content
Merck
All Photos(1)

Key Documents

45796

Supelco

2-Methylnaphthalene

analytical standard

Synonym(s):

β-Methylnaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H10
CAS Number:
Molecular Weight:
142.20
Beilstein:
906859
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Cc1ccc2ccccc2c1

InChI

1S/C11H10/c1-9-6-7-10-4-2-3-5-11(10)8-9/h2-8H,1H3

InChI key

QIMMUPPBPVKWKM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Methylnaphthalene belongs to the class of polycyclic aromatic hydrocarbons (PAHs), found to be a major environmental contaminant. It can occur in high concentrations in a variety of commercial products including coal tar, crude oil, etc.

Application

2-Methylnaphthalene may be used to identify the analyte from thermal degradation of bituminous/anthracite coals, tyres, and cellulose samples using gas chromatography-mass spectrometry (GC-MS) technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluating emission levels of polycyclic aromatic hydrocarbons from organic materials by analytical pyrolysis
Fabbri D and Vassura I
Journal of Analytical and Applied Pyrolysis, 75(2), 150-158 (2006)
Anaerobic degradation of 2-methylnaphthalene by a sulfate-reducing enrichment culture
Annweiler E, et al.
Applied and Environmental Microbiology, 66(12), 5329-5333 (2000)
Degradation of 2-methylnaphthalene by a sulfate-reducing enrichment culture of mesophilic freshwater bacteria
Galushko SA, et al.
Polycyclic Aromat. Compd., 23(2), 207-218 (2003)
Michael Safinowski et al.
Environmental microbiology, 8(2), 347-352 (2006-01-21)
The sulfate-reducing culture N47 can utilize naphthalene or 2-methylnaphthalene as the sole carbon source and electron donor. Here we show that the initial reaction in the naphthalene degradation pathway is a methylation to 2-methylnaphthalene which then undergoes the subsequent oxidation
Radosław Swiercz et al.
International journal of occupational medicine and environmental health, 23(4), 385-389 (2011-02-11)
The aim of the study was to evaluate the toxicokinetics of 2-methylnaphtalene (2-MN) during and after inhalation exposure. Male Wistar rats were exposed to 2-MN vapours at nominal concentrations of 200 or 400 mg/m3 in the dynamic inhalation chamber for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service