Skip to Content
Merck
All Photos(1)

Key Documents

45753

Supelco

Flusilazole

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H15F2N3Si
CAS Number:
Molecular Weight:
315.39
Beilstein:
5824097
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

C[Si](Cn1cncn1)(c2ccc(F)cc2)c3ccc(F)cc3

InChI

1S/C16H15F2N3Si/c1-22(12-21-11-19-10-20-21,15-6-2-13(17)3-7-15)16-8-4-14(18)5-9-16/h2-11H,12H2,1H3

InChI key

FQKUGOMFVDPBIZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flusilazole is an organosilicon fungicide, which belongs to the class of sterol demethylation inhibitors. Its mode of action involves the inhibition of lanosterol 1,4-α-demethylation in fungal sterol biosynthesis.

Application

Flusilazole may be used as a reference standard for the determination of flusilazole in water samples using solid-phase extraction (SPE) method using multi-walled carbon nanotubes as adsorbent coupled with high-performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Determination of pyrazole and pyrrole pesticides in environmental water samples by solid-phase extraction using multi-walled carbon nanotubes as adsorbent coupled with high-performance liquid chromatography.
Ma J, et al.
Journal of Chromatographic Science, 53(2), 380-384 (2014)
Sensitivity of Venturia inaequalis isolates from British Columbia to flusilazole and myclobutanil.
Sholberg LP and Haag DP
Canadian Journal of Plant Pathology, 15(2), 102-106 (1993)
J M Trzaskos et al.
Antimicrobial agents and chemotherapy, 33(8), 1228-1231 (1989-08-01)
Flusilazole inhibits the 14 alpha-demethylation of [24,25-3H-2]dihydrolanosterol by yeast and rat liver cell-free preparations. The 50% inhibitory concentration of demethylation shows that the yeast system is 100 times more sensitive than the rat system. Purified rat liver P-45014DM is about
Dorien A M van Dartel et al.
Toxicology and applied pharmacology, 251(2), 110-118 (2011-01-05)
The murine embryonic stem cell test (EST) is designed to evaluate developmental toxicity based on compound-induced inhibition of embryonic stem cell (ESC) differentiation into cardiomyocytes. The addition of transcriptomic evaluation within the EST may result in enhanced predictability and improved
Chen Wang et al.
Environmental monitoring and assessment, 185(11), 9169-9176 (2013-05-10)
In this paper, dissipation dynamic and terminal residue of flusilazole in mandarin and soil, as well as residue distribution of flusilazole in mandarin, were studied at three sites in China. Mandarin peel, mandarin pulp, whole mandarin, and soil samples were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service