Skip to Content
Merck
All Photos(1)

Key Documents

45701

Supelco

Triforin

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H14Cl6N4O2
CAS Number:
Molecular Weight:
434.96
Beilstein:
626358
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

ClC(Cl)(Cl)C(NC=O)N1CCN(CC1)C(NC=O)C(Cl)(Cl)Cl

InChI

1S/C10H14Cl6N4O2/c11-9(12,13)7(17-5-21)19-1-2-20(4-3-19)8(18-6-22)10(14,15)16/h5-8H,1-4H2,(H,17,21)(H,18,22)

InChI key

RROQIUMZODEXOR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triforin is a systemic fungicide, commonly used for the control of scab, petal blight, brown rot, black spot, rust and other diseases affecting fruits and vegetables.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Triforin may be used as an analytical reference standard for the quantification of the analyte in food commodities using different chromatography techniques.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sittig's Handbook of Pesticides and Agricultural Chemicals, 11(2), 506-513 (2014)
Method validation for the analysis of pesticide residues in grain by thin-layer chromatography
Tiryaki.O
Accreditation and Quality Assurance, 11(10), 506-513 (2006)
V N Golubev et al.
Biofizika, 34(1), 42-44 (1989-01-01)
It is found that bilayer lipid membranes acquire little cationic selectivity in the presence of systemic fungicide triforine at physiological pH, and besides potassium selectivity exceeds the sodium one. A decrease of pH to 3.5 leads to substitution of cationic
A T Wojdyła
Mededelingen (Rijksuniversiteit te Gent. Fakulteit van de Landbouwkundige en Toegepaste Biologische Wetenschappen), 66(2a), 167-177 (2002-11-12)
Efficacy of grapefruit extract (a.i. of Biosept 33 SL) in the control of Sphaerotheca pannosa var. rosae and Diplocarpon rosae on roses was investigated during 1998-1999. The extract was applied as plant spray in concentrations from 0.017 to 0.099%. First
[Determination of triforin in water and soil by a thin-layer chromatographic method].
P Iu Iurgaĭtene et al.
Gigiena i sanitariia, (1)(1), 46-47 (1981-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service