Skip to Content
Merck
All Photos(2)

Key Documents

45687

Supelco

Thiobencarb

PESTANAL®, analytical standard

Synonym(s):

Benthiocarb

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16ClNOS
CAS Number:
Molecular Weight:
257.78
Beilstein:
1968440
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCN(CC)C(=O)SCc1ccc(Cl)cc1

InChI

1S/C12H16ClNOS/c1-3-14(4-2)12(15)16-9-10-5-7-11(13)8-6-10/h5-8H,3-4,9H2,1-2H3

InChI key

QHTQREMOGMZHJV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Thiobencarb is a thiocarbamate herbicide, widely used for the control of pests in rice fields.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Thiobencarb may be used as a reference standard for the determination of thiobencarb residues in fruits and vegetables by matrix solid-phase dispersion and liquid chromatography coupled with mass spectrometry.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongbin Wei et al.
Chemosphere, 72(9), 1303-1308 (2008-06-07)
In order to integratedly evaluate the biological safety as a water quality index, an assessment method based on three toxicity tests (algae growth inhibition, daphnia immobilization and larval fish toxicity) was developed. In this study, the developed method was used
Amrith S Gunasekara et al.
Journal of agricultural and food chemistry, 53(26), 10113-10119 (2005-12-22)
The potential for reductive dechlorination of the herbicide thiobencarb (TB) by microbes and its prevention in saturated anaerobic rice field soils was examined in laboratory microcosms. TB is effective in controlling both annual grasses and broadleaf weeds. In anoxic microcosms
Sandy Raimondo et al.
Environmental toxicology and chemistry, 24(3), 564-572 (2005-03-23)
Population-level effects of the mysid, Americamysis bahia, exposed to varying thiobencarb concentrations were estimated using stage-structured matrix models. A deterministic density-independent matrix model estimated the decrease in population growth rate (lambda) with increasing thiobencarb concentration. An elasticity analysis determined that
Mojtaba Mahmoudi et al.
Chemosphere, 91(7), 1009-1017 (2013-03-07)
Herbicides are an important source of contamination in paddy fields. Monitoring their fate and chemical interactions is therefore imperative for sustaining the environment and human health. To meet this purpose, field experiments were conducted to investigate kinetics of thiobencarb and
Md Nurul Amin et al.
Chemosphere, 70(3), 511-515 (2007-10-30)
A cost-effective method with zero valent iron (ZVI) powder was developed for the purification of thiobencarb (TB)-contaminated water. The removal treatment was performed in the batch system. A sample solution of 10 ml containing 10 microg ml(-1) of TB could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service