Skip to Content
Merck
All Photos(1)

Key Documents

45667

Supelco

2,4,5-T

PESTANAL®

Synonym(s):

2,4,5-Trichlorophenoxyacetic acid, 2,4,5-T

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2OCH2CO2H
CAS Number:
Molecular Weight:
255.48
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

product line

PESTANAL®

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

154-158 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)COc1cc(Cl)c(Cl)cc1Cl

InChI

1S/C8H5Cl3O3/c9-4-1-6(11)7(2-5(4)10)14-3-8(12)13/h1-2H,3H2,(H,12,13)

InChI key

SMYMJHWAQXWPDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Anh Duc Ngo et al.
European journal of epidemiology, 25(1), 37-44 (2009-11-07)
The objective of this study is to conduct a meta-analysis of published and unpublished studies that examine the association between Agent Orange (AO) exposure and the risk of spina bifida. Relevant studies were identified through a computerized literature search of
Maria P Yurkova et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 12(4), 684-689 (2012-10-23)
Photodegradation of herbicide 2,4,5-trichlorophenoxyacetic acid (2,4,5-T) in aqueous solution was investigated by stationary (254 nm) and nanosecond laser (266 nm) flash photolysis techniques. It was shown that in the primary photochemical step both photoionization (which generates a hydrated electron-radical cation
Alvin L Young et al.
Military medicine, 176(7 Suppl), 29-34 (2011-09-16)
Serum dioxin studies of Vietnam (VN) veterans, military historical records of tactical herbicide use in Vietnam, and the compelling evidence of the photodegradation of 2,3,7,8-tetrachlorodibenzo-p-dioxin (TCDD) and other aspects of environmental fate and low bioavailability of TCDD are consistent with
Representations and reproductive hazards of Agent Orange.
Leslie J Reagan
The Journal of law, medicine & ethics : a journal of the American Society of Law, Medicine & Ethics, 39(1), 54-61 (2011-02-15)
Letter to the editor and response. Re: Production of illicit drugs, the environment, and human health, J of Toxicology and Environmental Medicine 2009;72 (15-16).
Jaime G Gomez
Journal of toxicology and environmental health. Part A, 73(9), 581-581 (2010-04-15)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service