Skip to Content
Merck
All Photos(1)

Key Documents

45526

Supelco

Hexachlorophene

PESTANAL®, analytical standard

Synonym(s):

2,2′-Methylenebis(3,4,6-trichlorophenol)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2[C6H(Cl)3OH]2
CAS Number:
Molecular Weight:
406.90
Beilstein:
2064407
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

163-165 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Oc1c(Cl)cc(Cl)c(Cl)c1Cc2c(O)c(Cl)cc(Cl)c2Cl

InChI

1S/C13H6Cl6O2/c14-6-2-8(16)12(20)4(10(6)18)1-5-11(19)7(15)3-9(17)13(5)21/h2-3,20-21H,1H2

InChI key

ACGUYXCXAPNIKK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seoyoung Park et al.
Molecular pharmacology, 70(3), 960-966 (2006-06-01)
Aberrant activation of Wnt/beta-catenin signaling and subsequent up-regulation of beta-catenin response transcription (CRT) is a critical event in the development of human colon cancer. Thus, Wnt/beta-catenin signaling is an attractive target for the development of anticancer therapeutics. In this study
J P Hachem et al.
Dermatology (Basel, Switzerland), 202(4), 314-319 (2001-07-17)
Nickel (Ni) allergic contact dermatitis (ACD) alters the skin barrier. Our aim was to compare the efficacy of combination therapies on ACD, using a topical corticosteroid and a corneotherapy agent (barrier cream), with that of a single therapy with corticosteroids.
Bernd Hutter et al.
Antimicrobial agents and chemotherapy, 48(8), 2838-2844 (2004-07-27)
We have generated a database of expression profiles carrying the transcriptional responses of the model organism Bacillus subtilis following treatment with 37 well-characterized antibacterial compounds of different classes. The database was used to build a predictor for the assignment of
Hye-Jin Min et al.
Cancer letters, 276(2), 136-142 (2008-12-19)
Many studies have shown that the activation of beta-catenin signaling can promote oncogenesis, and it is therefore of interest to find agents that modulate this pathway. Recent work has shown using B lymphoma cells that infection by Epstein-Barr virus (EBV)
Dao M Nguyen et al.
Emerging infectious diseases, 11(4), 526-532 (2005-04-15)
An outbreak of community-associated methicillin-resistant Staphylococcus aureus (MRSA) skin and soft tissue infection (SSTI) occurred in a college football team from August to September 2003. Eleven case-players were identified, and boils were the most common sign. Linemen had the highest

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service