Skip to Content
Merck
All Photos(1)

Key Documents

45473

Supelco

Ethephon

PESTANAL®, analytical standard

Synonym(s):

2-Chloroethylphosphonic acid, Ethephon

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2P(O)(OH)2
CAS Number:
Molecular Weight:
144.49
Beilstein:
1751208
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

70-72 °C (lit.)

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

OP(O)(=O)CCCl

InChI

1S/C2H6ClO3P/c3-1-2-7(4,5)6/h1-2H2,(H2,4,5,6)

InChI key

UDPGUMQDCGORJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1C

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsien-Jung Chen et al.
Journal of plant physiology, 169(1), 86-97 (2011-09-07)
In this report a full-length cDNA, SPCAT1, was isolated from ethephon-treated mature L3 leaves of sweet potato. SPCAT1 contained 1479 nucleotides (492 amino acids) in its open reading frame, and exhibited high amino acid sequence identities (ca. 71.2-80.9%) with several
Shigemi Seo et al.
Plant physiology, 155(1), 502-514 (2010-11-16)
Rice (Oryza sativa) plants carrying the Pi-i resistance gene to blast fungus Magnaporthe oryzae restrict invaded fungus in infected tissue via hypersensitive reaction or response (HR), which is accompanied by rapid ethylene production and formation of small HR lesions. Ethylene
Young Ock Ahn et al.
Molecular biology reports, 39(3), 2059-2067 (2011-06-07)
The expression profiles of three Brassica rapa metallothionein genes (BrMT 1-3) were determined in 7-day-old seedlings exposed to various exogenous factors including plant hormones, heavy metals and abiotic stresses. BrMT1, BrMT2, and BrMT3 were representatives of MT gene type 1
Hsien-Jung Chen et al.
Journal of plant physiology, 169(5), 529-541 (2012-01-10)
In this report a full-length cDNA, SPCAM, was isolated from ethephon-treated mature leaves of sweet potato. SPCAM contained 450 nucleotides (149 amino acids) in its open reading frame, and exhibited high amino acid sequence identities (ca. 76-100%) with several plant
Enriqueta Moyano-Cañete et al.
Plant & cell physiology, 54(2), 218-236 (2012-12-13)
Numerous GAST-like genes have been reported in higher plants, but only one GAST-like gene (FaGAST1) has been described in strawberry so far. Herein, we have identified a novel strawberry FaGAST gene (FaGAST2) whose expression showed an increase throughout fruit receptacle

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service