Skip to Content
Merck
All Photos(1)

Key Documents

45408

Supelco

Cycloate

PESTANAL®, analytical standard

Synonym(s):

S-Ethyl N-cyclohexyl-N-ethylthiocarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H21NOS
CAS Number:
Molecular Weight:
215.36
Beilstein:
2937178
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCSC(=O)N(CC)C1CCCCC1

InChI

1S/C11H21NOS/c1-3-12(11(13)14-4-2)10-8-6-5-7-9-10/h10H,3-9H2,1-2H3

InChI key

DFCAFRGABIXSDS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Aquatic Chronic 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Berta N Estevinho et al.
Journal of hazardous materials, 338, 85-92 (2017-05-23)
Molinate is a thiocarbamate herbicide used in rice crop protection. As other pesticides, molinate is a recognized environmental pollutant and bio-accumulated by some wildlife forms. Gulosibacter molinativorax ON4
B Kosinkiewicz et al.
Acta microbiologica Polonica, 33(2), 111-117 (1984-01-01)
The bacteria from Arthrobacter genus isolated from sugar beet rhizosphere were found to produce gallic, protocatechuic, p-hydroxybenzoic, syryngic, vanillic, veratric acids, p-quinone and two unidentified phenolic compounds. The mixture of the bacterial phenolic compounds increased the phytotoxicity of Roneet, inhibiting
J Czapski et al.
Journal of environmental science and health. Part. B, Pesticides, food contaminants, and agricultural wastes, 18(4-5), 497-503 (1983-08-01)
The influence of cycloate (active component of Ro-Neet 6E) on the biological value and residues in spinach was studied. During the growing season of spinach, cycloate residues decreased rapidly until by harvest time levels 0.0005--0.0008 mg/kg. had been reached. None
T P Suurmeijer et al.
Epilepsia, 42(9), 1160-1168 (2001-10-03)
Part of our research intended to explain "Quality of Life" (QoL) differences between people with epilepsy. To this end, a series of already existing generic and disease-specific health status measures were used. In this study, they were considered as determinants
Elzbieta G Magnucka et al.
Pest management science, 65(10), 1065-1070 (2009-05-30)
Cycloate inhibits the biosynthesis of very-long-chain fatty acids, the essential constituents of plant waxes and suberin. Fatty acids also serve as precursors of aliphatic carbon chains in resorcinolic lipids, which play a fundamental role in the plant defence system against

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service