Skip to Content
Merck
All Photos(1)

Documents

45407

Supelco

Cyanazine

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H13ClN6
CAS Number:
Molecular Weight:
240.69
Beilstein:
615509
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable
solid phase extraction (SPE): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCNc1nc(Cl)nc(NC(C)(C)C#N)n1

InChI

1S/C9H13ClN6/c1-4-12-7-13-6(10)14-8(15-7)16-9(2,3)5-11/h4H2,1-3H3,(H2,12,13,14,15,16)

InChI key

MZZBPDKVEFVLFF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Standard for Supelco MIP SPE cartridges. For more information request Supelco Literature T407075, T706023

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Mohammad Bagher Gholivand et al.
Analytica chimica acta, 713, 36-44 (2011-12-28)
A computational approach was used for screening functional monomers and polymerization solvent in the rational design of molecularly imprinted polymers (MIPs). It was based on the comparison of the binding energy of the complexes between the template and functional monomers.
Sabine Beulke et al.
Journal of environmental quality, 34(6), 1933-1943 (2005-10-14)
There is evidence that degradation of pesticides in simple laboratory systems may differ from that in the field, but it is not clear which of the simplifications inherent in laboratory studies present serious shortcomings. Laboratory experiments evaluated several simplifying assumptions
Mary Anne Nelson et al.
Analytical chemistry, 76(3), 805-813 (2004-01-31)
A portable system based on immunoextraction and reversed-phase HPLC was developed for the field analysis of herbicides in groundwater and surface water. Atrazine, simazine, and cyanazine were used as model analytes for this work. These were measured in water by
N C Hansen et al.
Journal of environmental quality, 30(6), 2120-2126 (2002-01-16)
Herbicides transported to surface waters by agricultural runoff are partitioned between solution and solid phases. Conservation tillage that reduces upland erosion will also reduce transport of herbicides associated with the solid phase. However, transport of many herbicides occurs predominantly in
Shannon M Lynch et al.
Environmental health perspectives, 114(8), 1248-1252 (2006-08-03)
Cyanazine is a common pesticide used frequently in the United States during the 1980s and 1990s. Animal and human studies have suggested that triazines may be carcinogenic, but results have been mixed. We evaluated cancer incidence in cyanazine-exposed pesticide applicators

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service