Skip to Content
Merck
All Photos(1)

Key Documents

45379

Supelco

Chlordecone

PESTANAL®, analytical standard

Synonym(s):

Kepone, Chlordecone, Perchloropentacyclo[5.3.0.02,6.03,9.04,8]decan-5-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10Cl10O
CAS Number:
Molecular Weight:
490.64
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

ClC1(Cl)C2(Cl)C3(Cl)C4(Cl)C(=O)C5(Cl)C3(Cl)C1(Cl)C5(Cl)C24Cl

InChI

1S/C10Cl10O/c11-2-1(21)3(12)6(15)4(2,13)8(17)5(2,14)7(3,16)9(6,18)10(8,19)20

InChI key

LHHGDZSESBACKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chlordecone is a chlorinated cyclodiene pesticide and a persistent environmental contaminant, obtained via degradation of mirex.

Application

Chlordecone may be used as an analytical reference standard for the quantification of the analyte in animals using different chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toxicological Study of Pesticides in Animals (1990)
Development and validation of an HPLC-MS/MS method with QuEChERS extraction using isotopic dilution to simultaneously analyze chlordecone and chlordecol in animal livers
Saint-Hilaire M, et al.
Food Chemistry, 252, 147-153 (2018)
High-performance liquid chromatography separation of chlordecone and its metabolites
Chromatographia, 22(1-6), 165-167 (1986)
Fei Wang et al.
International immunopharmacology, 7(13), 1808-1812 (2007-11-13)
In murine models of systemic lupus erythematosus (SLE), administration of either prolactin or estradiol (E2) increases autoimmunity, and there is evidence that elevated prolactin in response to E2 administration may contribute substantially to E2 effects. Hormonal influence on SLE can
Didier Brunet et al.
Environmental pollution (Barking, Essex : 1987), 157(11), 3120-3125 (2009-06-06)
Chlordecone is a toxic organochlorine insecticide that was used in banana plantations until 1993 in the French West Indies. This study aimed at assessing the potential of near infrared reflectance spectroscopy (NIRS) for determining chlordecone content in Andosols, Nitisols and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service