Skip to Content
Merck

44514

Dowex 50W X8 Ion Exchange Resin

sodium form, 100-200 mesh, strongly acidic

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Dowex 50WX8, Na+-form, strongly acidic, 100-200 mesh

form

beads

crosslinking

8 % cross-linked

technique(s)

LPLC: suitable

loss

~45% loss on drying, 110 °C

matrix

styrene-divinylbenzene

matrix active group

sulfonic acid functional group

particle size

100-200 mesh

capacity

1.7 meq/mL

separation technique

cation exchange

SMILES string

[Na+].[C-H2]Cc3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H11.C10H10.C8H8.Na/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;/h3,5-8H,1-2,4H2;3-8H,1-2H2;2-7H,1H2;/q-1;;;+1

InChI key

NEQWAZPDMBONCG-UHFFFAOYSA-N

Application


  • Heterogeneous Brønsted Catalysis in the Solvent-Free and Multigram-Scale Synthesis of Polyalcohol Acrylates: The Case Study of Trimethylolpropane Triacrylate.: This research demonstrates the application of Dowex 50WX8 as a catalyst in the eco-friendly synthesis of polyalcohol acrylates, highlighting its potential to facilitate sustainable chemical processes in industrial settings (Melchiorre et al., 2024).

  • Chiral separation of propranolol by electrokinetic chromatography using nanodiamonds and human serum albumin as a pseudo-stationary phase in river water.: Explores the use of Dowex 50WX8 in chiral separations, contributing to the environmental analysis of pharmaceutical contaminants and enhancing understanding of chiral drug behavior in aquatic environments (Lanaro et al., 2024).

  • Modeling and equilibrium studies on the recovery of praseodymium (III), dysprosium (III) and yttrium (III) using acidic cation exchange resin.: Discusses the efficiency of Dowex 50WX8 in recovering valuable rare earth elements from waste streams, offering insights into resource recovery and environmental sustainability (Masry et al., 2022).

  • Improved separation scheme for (44)Sc produced by irradiation of (nat)Ca targets with 12.8 MeV protons.: Illustrates the role of Dowex 50WX8 in the radiochemical separation of scandium isotopes, crucial for medical imaging and therapy applications, showcasing the resin′s capability in nuclear medicine (Kurakina et al., 2022).

  • Supplementary data on rapid sample clean-up procedure of aminophosphonates for LC/MS analysis.: Highlights the use of Dowex 50WX8 in the pre-analysis clean-up steps for liquid chromatography-mass spectrometry, improving the reliability and efficiency of analytical methods in biochemical research (Kuhn et al., 2020).

Legal Information



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library