Skip to Content
Merck
All Photos(1)

Key Documents

43861

Supelco

Furan

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4O
CAS Number:
Molecular Weight:
68.07
Beilstein:
103221
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

vapor density

2.35 (vs air)

vapor pressure

1672 mmHg ( 55 °C)
31.66 psi ( 55 °C)
493 mmHg ( 20 °C)
9.22 psi ( 20 °C)

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

contains

~0.025% 2,6-di-tert-4-methylphenol as stabilizer

expl. lim.

14.3 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.421 (lit.)
n20/D 1.421

bp

32 °C/758 mmHg (lit.)

density

0.936 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

c1ccoc1

InChI

1S/C4H4O/c1-2-4-5-3-1/h1-4H

InChI key

YLQBMQCUIZJEEH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Furan is a clear and colorless liquid, and a bimolecular heterocycle, which can be used to trap unstable intermediates containing highly reactive species, via Diels-Alder cycloaddition reactions.[1]
Furan, a naturally occurring compound associated with the flavor of food, is found at very low levels in a variety of food products and beverages. It finds applications as a solvent for resins and lacquers as well as in the preparation of organic compounds, in pharmaceuticals, etc. Furan is also reportedly identified in heat treated food products, including cooked chicken, fish protein concentrate, caramel back, canned meat, bread, coffee, etc.[2]

Application

Furan can undergo Diels-Alder [4+2] cycloaddition reaction with maleic anhydride to form the corresponding exo and endo furan-maleic anhydride adducts.[1]
Furan may be used as an analytical reference standard for the quantification of the analyte in heat-processed foods using gas chromatography-mass spectrometry (GC-MS).[3][4]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 1 - Muta. 2 - Skin Irrit. 2 - STOT RE 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-32.8 °F - closed cup

Flash Point(C)

-36 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of furan in heat-processed foods in China by automated headspace gas chromatography-mass spectrometry (HS-GC-MS)
Nie P-S, et al.
Food Control, 30(1), 62-68 (2013)
Furan in Food ? a Review
Vranova J and Ciesarova Z
Czech Journal of Food Sciences, 27(1), 1-10 (2009)
Synthetic applications of furan Diels-Alder chemistry
Kappe O.C, et al.
Tetrahedron, 53(42), 14179-14233 (1997)
Simultaneous determination of furan and 2-alkylfurans in heat-processed foods by automated static headspace gas chromatography-mass spectrometry
Shen M, et al.
LWT--Food Science and Technology, 72, 44-54 (2016)
Prashant Sonar et al.
Chemical communications (Cambridge, England), 49(16), 1588-1590 (2012-11-09)
A fluorenone based alternating copolymer () with a furan based fused aromatic moiety has been designed and synthesized. exhibits a small band gap with a lower HOMO value. Testing this polymer semiconductor as the active layer in organic thin-film transistors

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service