Skip to Content
Merck
All Photos(3)

Key Documents

434760

Sigma-Aldrich

Benzamidine hydrochloride

99%

Synonym(s):

Amidinobenzene hydrochloride, Benzamidinium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
156.61
Beilstein:
3594959
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

99%

form

beads

mp

169-173 °C (lit.)

solubility

water: soluble

SMILES string

Cl.NC(=N)c1ccccc1

InChI

1S/C7H8N2.ClH/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H3,8,9);1H

InChI key

LZCZIHQBSCVGRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

<ul>
<li><strong>Purification and characterization of a serine protease from organic dust and elucidation of its inductive effects on lung i</strong></li>
<li><strong>Preclinical testing of dabigatran in trypsin-dependent pancreatitis:</strong> This research uses Benzamidine hydrochloride to inhibit serine proteases in models of pancreatitis, highlighting its potential in preclinical studies to evaluate new therapeutic approaches for inflammatory and digestive disorders (Pesei et al., 2022).</li>
</ul>

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seweryn Mroczek et al.
Nature communications, 8(1), 619-619 (2017-09-22)
FAM46C is one of the most frequently mutated genes in multiple myeloma. Here, using a combination of in vitro and in vivo approaches, we demonstrate that FAM46C encodes an active non-canonical poly(A) polymerase which enhances mRNA stability and gene expression.
Joris Van Lindt et al.
Communications biology, 4(1), 77-77 (2021-01-21)
Understanding the kinetics, thermodynamics, and molecular mechanisms of liquid-liquid phase separation (LLPS) is of paramount importance in cell biology, requiring reproducible methods for studying often severely aggregation-prone proteins. Frequently applied approaches for inducing LLPS, such as dilution of the protein
Anita de Ruiter et al.
Journal of computational chemistry, 34(12), 1024-1034 (2013-01-22)
The performances of Bennett's acceptance ratio method and thermodynamic integration (TI) for the calculation of free energy differences in protein simulations are compared. For the latter, the standard trapezoidal rule, Simpson's rule, and Clenshaw-Curtis integration are used as numerical integration
Zaofeng Yang et al.
iScience, 23(6), 101158-101158 (2020-05-29)
The effectiveness of cancer radiotherapy is frequently hindered by the hypoxia of the tumor microenvironment. Direct delivery of oxygen to hypoxic tumor tissues is an attractive strategy to overcome this hypoxia-associated radioresistance. Herein, we report the generation of submicron-sized particles
Bradley S Heater et al.
Biotechnology for biofuels, 12, 165-165 (2019-07-13)
We have recently developed a one-step, genetically encoded immobilization approach based on fusion of a target enzyme to the self-crystallizing protein Cry3Aa, followed by direct production and isolation of the fusion crystals from Bacillus thuringiensis. Using this approach, Bacillus subtilis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service