Skip to Content
Merck

41349

Amberlite FPC23 Ion Exchange Resin

greener alternative

hydrogen form, 16-40 mesh, strongly acidic

Synonym(s):

Amberlite FPC23 H

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116105
NACRES:
SB.52
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Amberlite FPC23, hydrogen form, strongly acidic, 16-40 mesh

Quality Segment

form

beads

greener alternative product characteristics

Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

LPLC: suitable

surface area

800 m2/g

loss

40-55% loss on drying

matrix

styrene-divinylbenzene

matrix active group

polymer

particle size

16-40 mesh

pore size

200 Å mean pore size

greener alternative category

separation technique

reversed phase

SMILES string

[S](=O)(=O)(O)c2c(cccc2)C=C.c1(c(cccc1)C=C)C=C

InChI

1S/C10H10.C8H8O3S/c1-3-9-7-5-6-8-10(9)4-2;1-2-7-5-3-4-6-8(7)12(9,10)11/h3-8H,1-2H2;2-6H,1H2,(H,9,10,11)

InChI key

SIWVGXQOXWGJCI-UHFFFAOYSA-N

General description

Amberlite FPC23 is proton ion-exchanger resin. Amberlite FPC23 H, macroreticular strongly acidic cation exchange resin for deashing of corn syrups and the softening of beet sugar syrups.

Application

Amberlite FPC23 was used to obtain protonated nanostructured aluminosilicate through ion-exchange columns.

Features and Benefits

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product has been enhanced for catalytic efficiency. Click here for more information.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library