Skip to Content
Merck

Skip To

40772

3-(N,N-Dimethylmyristylammonio)propanesulfonate

≥98.0% (TLC)

Synonym(s):

3-(N,N-Dimethyltetradecylammonio)propanesulfonate, 3-(Myristyldimethylammonio)propanesulfonate, N-Tetradecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate, Myristyl sulfobetaine, SB3-14

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)13N+(CH3)2CH2CH2CH2SO3-
CAS Number:
Molecular Weight:
363.60
UNSPSC Code:
12161900
NACRES:
NA.25
PubChem Substance ID:
EC Number:
239-003-9
Beilstein/REAXYS Number:
6419727
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

Quality Segment

assay

≥98.0% (TLC)

form

powder

mol wt

micellar avg mol wt 30,200

aggregation number

83

technique(s)

nucleic acid detection: suitable, protein purification: suitable, protein quantification: suitable

CMC

0.1-0.4 mM (20-25°C)

mp

246 - 248  °C

solubility

water: 100 g/L at 20 °C

SMILES string

CCCCCCCCCCCCCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C19H41NO3S/c1-4-5-6-7-8-9-10-11-12-13-14-15-17-20(2,3)18-16-19-24(21,22)23/h4-19H2,1-3H3

InChI key

BHATUINFZWUDIX-UHFFFAOYSA-N

General description

Zwitterionic detergent.

Other Notes

Zwitterionic surfactant which effectively solubilises cardiac 5′-nucleotidase[1]; Micorsomal proteins; H+-ATPase from corn roots

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T0807T7763H6883
assay

≥98.0% (TLC)

assay

≥99% (TLC)

assay

≥99% (TLC)

assay

≥98% (TLC)

form

powder

form

-

form

-

form

-

technique(s)

nucleic acid detection: suitable, protein quantification: suitable, protein purification: suitable

technique(s)

electrophoresis: suitable, ion chromatography: suitable

technique(s)

electrophoresis: suitable

technique(s)

electrophoresis: suitable, mass spectrometry (MS): suitable, thin layer chromatography (TLC): suitable

aggregation number

83

aggregation number

83

aggregation number

83

aggregation number

155

mol wt

micellar avg mol wt 30,200

mol wt

micellar avg mol wt 30,200

mol wt

micellar avg mol wt 30,200

mol wt

micellar avg mol wt 60,700

CMC

0.1-0.4 mM (20-25°C)

CMC

0.1-0.4 mM (20-25°C)

CMC

0.1-0.4 mM (20-25°C)

CMC

0.01-0.06 mM (20-25°C)


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters