Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
C6H4[COO(CH2)9CH3]2
CAS Number:
Molecular Weight:
446.66
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
201-561-6
Beilstein/REAXYS Number:
1893077
MDL number:
grade
analytical standard
Quality Level
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
SMILES string
CCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCCCCC
InChI
1S/C28H46O4/c1-3-5-7-9-11-13-15-19-23-31-27(29)25-21-17-18-22-26(25)28(30)32-24-20-16-14-12-10-8-6-4-2/h17-18,21-22H,3-16,19-20,23-24H2,1-2H3
InChI key
PGIBJVOPLXHHGS-UHFFFAOYSA-N
General description
Didecyl phthalate belongs to the class of phthalate esters widely used as plasticizers in plastic films.
Application
Didecyl phthalate has been used as stationary liquid phase in the analysis and separation of terpene hydrocarbons and other related compounds using gas chromatography method (GC).
Didecyl phthalate may be used as an analytical reference standard for the quantification of the analyte in environmental samples, plastics and agricultural plastic films using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
392.0 °F - closed cup
flash_point_c
200 °C - closed cup
ppe
Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Analysis of terpene hydrocarbons and related compounds by gas chromatography
Zubyk.J.K and Conner.Z.A
Analytical Chemistry, 32, 912-917 (1960)
Identification and determination of phthalate esters in river water by high-performance liquid chromatography
Mori S, et al.
Journal of Chromatography A, 129, 53-60 (1976)
Thermally stable ionic liquid-based sol--gel coating for ultrasonic extraction-solid-phase microextraction-gas chromatography determination of phthalate esters in agricultural plastic films
Zhou X, et al.
Talanta, 89, 129-135 (2012)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 36933-250MG | 04061831833072 |
| 36933-1EA | 04061831833065 |