Skip to Content
Merck
All Photos(1)

Key Documents

36908

Supelco

Dicyclohexyl phthalate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2C6H11)2
CAS Number:
Molecular Weight:
330.42
Beilstein:
1889288
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

11.6 (vs air)

vapor pressure

0.1 mmHg ( 150 °C)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

63-67 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

O=C(OC1CCCCC1)c2ccccc2C(=O)OC3CCCCC3

InChI

1S/C20H26O4/c21-19(23-15-9-3-1-4-10-15)17-13-7-8-14-18(17)20(22)24-16-11-5-2-6-12-16/h7-8,13-16H,1-6,9-12H2

InChI key

VOWAEIGWURALJQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dicyclohexyl phthalate is a commonly used plasticizer ingredient, which can be used in the production of vinyl acetate, nitrocellulose, ethyl cellulose, polyvinyl chloride etc.

Application

Dicyclohexyl phthalate may be used as an analytical reference standard for the determination of the analyte in cosmetic products, environmental samples, and sewage sludge samples by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

356.0 - 374.0 °F

Flash Point(C)

180 - 190 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of in utero through lactational exposure to dicyclohexyl phthalate and p, p?-DDE in Sprague?Dawley rats
Yamasaki K, et al.
Toxicology Letters, 189, 14-20 (2009)
A two-generation reproductive toxicity study of dicyclohexyl phthalate in rats
Hoshino N, et al.
The Journal of Toxicological Sciences, 30, S79-S96 (2005)
Trace determination of phthalate esters in river water by solvent sublation followed by high-performance liquid chromatography-ultraviolet detection
Guo L and Dong H
International Journal of Environmental Analytical Chemistry, 89(5), 357-365 (2009)
Determination of phthalates in crude extracts of sewage sludges by high-resolution capillary gas chromatography with mass spectrometric detection
Berset JD and Etter-Holzer R
Journal of AOAC (Association of Official Analytical Chemists) International, 84(2), 383-391 (2001)
D Asai et al.
Toxicology letters, 118(1-2), 1-8 (2001-01-04)
Xenoestrogen dialkyl phthalates, C(6)H(4)(COOC(n)H(m))(2), lack the phenolic hydroxyl group that is an essential structural component of the steroid A ring of 17 beta-estradiol. In order to examine whether dialkyl phthalates imitate the steroid structure, we have synthesized a series of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service