Skip to Content
Merck
All Photos(1)

Key Documents

36749

Supelco

2,4-Dichlorobenzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3CO2H
CAS Number:
Molecular Weight:
191.01
Beilstein:
1868192
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

157-160 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1ccc(Cl)cc1Cl

InChI

1S/C7H4Cl2O2/c8-4-1-2-5(7(10)11)6(9)3-4/h1-3H,(H,10,11)

InChI key

ATCRIUVQKHMXSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Ion Mobility: TWCCSN2 value of 146.1 Å2 [M-H]-

The collision cross section (CCS) measurement was provided by Waters Corporation, using the SYNAPT XS mass spectrometer.
For a description and overview of how ion mobility enables the measurement of the CCS of an ion visit ims.waters.com.
Further information on the SYNAPT XS mass spectrometer can be found on the IMS microsite and product webpage.

TWCCS measurements are expected to be within 2% of this reference value.

P/N 36749 is part of the Waters Extractables & Leachables UNIFI scientific library which can be downloaded from Waters Marketplace.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel J Repeta et al.
Environmental science & technology, 38(20), 5373-5378 (2004-11-17)
Chromophoric or colored dissolved organic matter (CDOM) is one of the principal light adsorbing components of seawater, particularly in the ultraviolet, where it attenuates over 90% of downwelling ultraviolet radiation. In highly productive coastal regions and throughout most of the
A Iu Sokolov et al.
Eksperimental'naia i klinicheskaia farmakologiia, 74(5), 13-16 (2011-08-04)
Neurophysiological experiments on anesthetized rats were used to study the effects of various doses (12.5, 25, 37.5 mg/kg, i.v.) of drug composition migrepin (representing a combination of potassium-2,4-dichlorobenzoate, carbamazepine, and caffeine) on background firing of the trigeminal nucleus caudalis neurons
Anthony D Wright et al.
Chemosphere, 65(4), 604-608 (2006-03-24)
The toxic effects of several species of fresh water cyanobacteria, notably Microcystis species and associated toxins, the microcystins, Anabaena species (anatoxin), Nodularia sp. (nodularin), and Cylindrospermopsis raciborskii (cylindrospermopsin), are well known. Little, however, is known about the effects of secondary
Sunday A Adebusoye et al.
Bioresource technology, 102(3), 3041-3048 (2010-11-16)
Multiple bacterial strains with CBA metabolic properties were isolated using a simple selective strategy. Phylogenetic analysis of the 16S rRNA gene sequences grouped them into two main clusters consisting of four bacterial phyla and belonging to 17 genera. Whereas growth
G M Zaitsev et al.
FEMS microbiology letters, 65(2), 171-176 (1991-06-15)
The strains of Arthrobacter globiformis KZT1, Corynebacterium sepedonicum KZ4 and Pseudomonas cepacia KZ2 capable of early dehalogenation and complete oxidation of 4-chloro-, 2,4-dichloro-and 2-chlorobenzoic acids, respectively, have been analyzed for the origin of the genetic control of degradation. The occurrence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service