Skip to Content
Merck
All Photos(2)

Key Documents

36704

Supelco

3,5-Dichloroaniline

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Beilstein:
636492
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

259-260 °C/741 mmHg (lit.)

mp

46-52 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1cc(Cl)cc(Cl)c1

InChI

1S/C6H5Cl2N/c7-4-1-5(8)3-6(9)2-4/h1-3H,9H2

InChI key

UQRLKWGPEVNVHT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Kidney

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

271.4 °F - closed cup

Flash Point(C)

133 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G O Rankin et al.
Toxicology letters, 30(2), 173-179 (1986-02-01)
The nephrotoxic potential of 3,5-dichloroaniline (DCA) was examined in male Sprague-Dawley rats. Rats were administered DCA (0.4, 0.8 or 1.0 mmol/kg, i.p.), or 0.9% saline (1.0 ml/kg, i.p.), and renal function was monitored at 24 and 48 h. DCA (0.4
Roberta Turci et al.
Rapid communications in mass spectrometry : RCM, 20(17), 2621-2625 (2006-08-10)
3,4- and 3,5-Dichloroanilines (DCAs) are common markers of some non-persistent pesticides, e.g. linuron, diuron, vinclozolin, and iprodione. The general population may be exposed to these DCAs and/or their precursors mainly through diet. Since adverse effects on human health, such as
Residue analysis of imazapyr and chlozolinate in water using sunlight.
A Santoro et al.
Bulletin of environmental contamination and toxicology, 63(1), 33-38 (1999-07-29)
Marbouh L El et al.
International journal of clinical pharmacology and therapeutics, 40(1), 41-46 (2002-02-12)
In order to develop a screening test to detect human occupational exposure to aromatic amines such as 3,4-dichloroaniline (3,4-DCA) and 3,5-dichloroaniline (3,5-DCA), we first investigated the urinary excretion of these highly toxic compounds in the rat. The study was performed
M A Valentovic et al.
Toxicology letters, 78(3), 207-214 (1995-08-01)
3,5-Dichloroaniline (3,5-DCA), a derivative needed in the manufacture of dyes, pesticides and industrial compounds has been reported to induce renal damage. This study investigated whether pretreatment with inducers or inhibitors of P450 altered 3,5-DCA toxicity. P450 levels were induced in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service