Skip to Content
Merck
All Photos(1)

Documents

36392

Supelco

Acetaldehyde solution

natural, 50 wt. % in ethanol, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CHO
CAS Number:
Molecular Weight:
44.05
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard
natural

Quality Level

Assay

≥98.0% (acetaldehyde & ethanol, GC)

shelf life

limited shelf life, expiry date on the label

concentration

≥48.0% (acetaldehyde, T)
50 wt. % in ethanol

refractive index

n20/D 1.380

density

0.868 g/mL at 20 °C

format

single component solution

storage temp.

2-8°C

SMILES string

[H]C(C)=O

InChI

1S/C2H4O/c1-2-3/h2H,1H3

InChI key

IKHGUXGNUITLKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetaldehyde comprises of a carbonyl group linked to a hydrogen atom. It is a well-known mutagen and carcinogen.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Acetaldehyde has been used as a reference standard for the determination of acetaldehyde as a volatile constituent of extra virgin olive oil (EVOO) by gas chromatography coupled with ion mobility spectrometry (IMS).
It may be used as a reference standard for the detection of acetaldehyde which is formed as a pyrolysis product of tobacco additives and as endogenous aldehyde in human urine by gas chromatography-mass spectrometry technique.

Analysis Note

acetaldehyde forms the diacetal in alcoholic solution

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Eye Irrit. 2 - Flam. Liq. 1 - Muta. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analysis of endogenous aldehydes in human urine by static headspace gas chromatography-mass spectrometry
Serrano, Maria and Gallego, Mercedes and Silva, Manuel
Journal of Chromatography A, 1437, 241-246 (2016)
Determination of volatile compounds by GC-IMS to assign the quality of virgin olive oil
Garrido-Delgado, Rocio and del Mar Dobao-Prieto, Maria and Arce, Lourdes and Valcarcel, Miguel
Food Chemistry, 187, 572-579 (2015)
Oxidative and inert pyrolysis on-line coupled to gas chromatography with mass spectrometric detection: On the pyrolysis products of tobacco additives
Paschke, Meike and Hutzler, Christoph and Henkler, Frank and Luch, Andreas
International Journal of Hygiene and Environmental Health, 219(8), 780-791 (2016)
Key role of local acetaldehyde in upper GI tract carcinogenesis
Salaspuro, Mikko
Best Practice & Research. Clinical Gastroenterology (2017)
Universities Press Dictionary Of Chem (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service