Skip to Content
Merck
All Photos(1)

Key Documents

36189

Supelco

Penconazol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H15Cl2N3
CAS Number:
Molecular Weight:
284.18
Beilstein:
541488
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCC(Cn1cncn1)c2ccc(Cl)cc2Cl

InChI

1S/C13H15Cl2N3/c1-2-3-10(7-18-9-16-8-17-18)12-5-4-11(14)6-13(12)15/h4-6,8-10H,2-3,7H2,1H3

InChI key

WKBPZYKAUNRMKP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Penconazol is a hydrophobic and systemic fungicide,[1] which is effectively used against powdery mildew in vineyards.[2]

Application

Penconazol may be used as a reference standard in the determination of penconazol in grapefruit samples using gas chromatography (GC).[3]
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David R Johnson et al.
Environmental microbiology, 11(7), 1682-1691 (2009-02-26)
Biocides are widely used for domestic hygiene, agricultural and industrial applications. Their widespread use has resulted in their introduction into the environment and raised concerns about potential deleterious effects on aquatic ecosystems. In this study, the toxicity of the biocides
M Goumenou et al.
Fresenius' journal of analytical chemistry, 370(7), 946-950 (2001-09-25)
A simple analytical method, validated in-house and intra-laboratory, has been developed for the determination of penconazole on personal protection equipment (PPE) used by operators during field applications of Topas 20EW. The analytical determination of penconazole was performed by GC-ECD. Sample
M S Rodríguez-Cruz et al.
Journal of hazardous materials, 160(1), 200-207 (2008-04-11)
Sorption-desorption of two different hydrophobic fungicides, penconazole and metalaxyl, by a series of clay minerals layered and non-layered (montmorillonite, illite, kaolinite, muscovite, sepiolite and palygorskite) modified with the two-chain cationic surfactant, dihexadecyldimetylammonium (DHDDMA) was studied by first time in this
Nicoleta A Suciu et al.
Environmental science and pollution research international, 18(8), 1374-1383 (2011-04-06)
The main objective of this work was to develop and test a pilot scheme for decontaminating pesticide-containing water derived from pesticide mixtures used to protect vineyards, in which the scheme comprises adsorption by an organoclay and includes a system where
Dalal Jawich et al.
Molecular nutrition & food research, 50(6), 552-556 (2006-05-27)
The aim of this study consisted to evaluate the impact of a pesticide (penconazole) on the growth kinetics and genotoxicity on two yeast strains (Saccharomyces cerevisiae and Metschnikowia pulcherrima). When the penconazole was added at different phases of the growth

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service