Skip to Content
Merck
All Photos(2)

Key Documents

35886

Supelco

Pentachlorobenzene

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC6Cl5
CAS Number:
Molecular Weight:
250.34
Beilstein:
1911550
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

275-277 °C (lit.)

mp

84-87 °C (lit.)

density

1.609 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1cc(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6HCl5/c7-2-1-3(8)5(10)6(11)4(2)9/h1H

InChI key

CEOCDNVZRAIOQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ewa Ł Gregoraszczuk et al.
Reproductive toxicology (Elmsford, N.Y.), 31(4), 494-499 (2011-02-08)
In the current study, we determined in vitro accumulation of hexachlorobenzene (HCBz) and pentachlorobenzene (PeCBz) in porcine ovarian follicles, the effect on steroidogenesis and the expression of enzymes responsible for steroid synthesis. Sixty percent of the HCBz and almost 100%
Alice Dvorska et al.
Environmental pollution (Barking, Essex : 1987), 156(2), 403-408 (2008-05-23)
The regional observatory Kosetice is a central European background station. Unique continuous monitoring from 1988 on is held here. POP (persistent organic pollutant) concentration values of air samples from Kosetice taken between 1996 and 2005 were statistically processed. Values of
Ian John Allan et al.
Journal of environmental monitoring : JEM, 13(9), 2420-2426 (2011-07-27)
Thirty two polychlorinated biphenyl congeners (PCBs), hexachlorobenzene (HCB) and pentachlorobenzene (PeCB) were analysed in passive sampler extracts from surface water-exposed semipermeable membrane devices (SPMDs) and in bed sediment samples from a small urban watercourse, the River Alna (Oslo, Norway). Performance
N Martí et al.
Marine pollution bulletin, 62(3), 615-625 (2011-02-08)
A comprehensive study aimed at evaluating the occurrence, significance of concentrations and spatial distribution of priority pollutants (PPs) along the Comunidad Valenciana coastal waters (Spain) was carried out in order to fulfil the European Water Framework Directive (WFD). Additionally, PP
Lance J Schuler et al.
Chemosphere, 77(3), 399-403 (2009-08-14)
Nonpolar organic chemicals such as polycyclic aromatic hydrocarbons and chlorobenzenes are expected to act additively when exposed as a mixture. The present study examined the toxicity of fluoranthene (FLU) and pentachlorobenzene (PCBz) individually and in a binary mixture using the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service