Skip to Content
Merck
All Photos(1)

Documents

35596

Supelco

PCB No 3

analytical standard

Synonym(s):

4-Chlorobiphenyl, 4-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9Cl
CAS Number:
Molecular Weight:
188.65
Beilstein:
774966
Ballschmiter Number:
3
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

neat

SMILES string

Clc1ccc(cc1)-c2ccccc2

InChI

1S/C12H9Cl/c13-12-8-6-11(7-9-12)10-4-2-1-3-5-10/h1-9H

InChI key

FPWNLURCHDRMHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polychlorinated biphenyls (PCBs) belong to the class of highly toxic pollutants commonly found in various environmental matrices and food products. They find a variety of applications as flame retardants, plasticizers, dielectric and heat transfer fluids, etc.

Application

PCB No 3 may be used as an analytical reference standard for the quantification of the analyte in aqueous solution in the presence of dissolved organic carbon using high-performance liquid chromatography technique.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Health hazardEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Markus Alexander Zettner et al.
Toxicological sciences : an official journal of the Society of Toxicology, 100(1), 88-98 (2007-08-10)
4-Monochlorobiphenyl (PCB3) is a component of commercial polychlorinated biphenyl (PCB) products and is an airborne environmental pollutant. Our recent study with transgenic Fischer 344 rats revealed the mutagenic potential of PCB3 in the livers of male rats. PCB3 is converted
Parvaneh Espandiari et al.
Toxicological sciences : an official journal of the Society of Toxicology, 79(1), 41-46 (2004-02-21)
We recently reported that several mono- to tetrachlorinated biphenyls have initiating activity in the livers of Fischer 344 rats. In the present study, we investigated the metabolic activation of one of those compounds, 4-chlorobiphenyl (PCB 3). Monohydroxy (400 micromol/kg), dihydroxy
Guangshu Zhai et al.
Environmental science & technology, 47(1), 557-562 (2012-12-12)
4-Monochlorobiphenyl (PCB3) has been proven to be transformed into hydroxylated metabolites of PCB3 (OH-PCB3s) in whole poplar plants in our previous work. However, hydroxylated metabolites of PCBs, including OH-PCB3s, as the substrates of sulfotransferases have not been studied in many
G Luthe et al.
Acta crystallographica. Section B, Structural science, 63(Pt 2), 319-327 (2007-03-22)
Accurate structure determinations by X-ray crystal analysis and computation using semi-empirical self-consistent field molecular orbital calculations are described and compared for five monofluorinated analogues of 4-chlorobiphenyl, i.e. 2-fluoro-4-chlorobiphenyl, 2'-fluoro-4-chlorobiphenyl, 3-fluoro-4-chlorobiphenyl, 3'-fluoro-4-chlorobiphenyl and 4'-fluoro-4-chlorobiphenyl. Intermolecular interactions for all monofluorinated isomers are
Guangshu Zhai et al.
Environmental science & technology, 44(10), 3901-3907 (2010-04-21)
4-Monochlorobiphenyl (CB3), mainly an airborne pollutant, undergoes rapid biotransformation to produce hydroxylated metabolites (OH-CB3s). However, up to now, hydroxylation of CB3 has not been studied in living organisms. In order to explore the formation of hydroxylated metabolites of CB3 in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service