34514
Malathion-(diethyl-d10)
PESTANAL®, analytical standard
Synonym(s):
S-[1,2-Bis(ethoxy-d5-carbonyl)ethyl] O,O-dimethyl phosphorodithioate
About This Item
Recommended Products
grade
analytical standard
Quality Level
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
application(s)
agriculture
environmental
format
neat
storage temp.
2-8°C
SMILES string
[P](=S)(SC(CC(=O)OC(C([2H])([2H])[2H])([2H])[2H])C(=O)OC(C([2H])([2H])[2H])([2H])[2H])(OC)OC
InChI
1S/C10H19O6PS2/c1-5-15-9(11)7-8(10(12)16-6-2)19-17(18,13-3)14-4/h8H,5-7H2,1-4H3/i1D3,2D3,5D2,6D2
InChI key
JXSJBGJIGXNWCI-JKSUIMTKSA-N
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service