Skip to Content
Merck
All Photos(1)

Key Documents

34078

Supelco

Oxyclozanide

VETRANAL®, analytical standard

Synonym(s):

2,3,5-Trichloro-N-(3,5-dichloro-2-hydroxyphenyl)-6-hydroxybenzamide, 3,3′,5,5′,6-Pentachloro-2′-hydroxysalicylanilide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H6Cl5NO3
CAS Number:
Molecular Weight:
401.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Oc1c(Cl)cc(Cl)cc1NC(=O)c2c(O)c(Cl)cc(Cl)c2Cl

InChI

1S/C13H6Cl5NO3/c14-4-1-6(16)11(20)8(2-4)19-13(22)9-10(18)5(15)3-7(17)12(9)21/h1-3,20-21H,(H,19,22)

InChI key

JYWIYHUXVMAGLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Oxyclozanide is an anthelmintic agent, which can show activity against Fasciola hepatica, a parasitic trematode and hence, can be used in the treatment of liver fluke infections.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

S A Spence et al.
Australian veterinary journal, 69(9), 217-220 (1992-09-01)
A significant increase in milk production, averaging 164 litres per cow per lactation (a 4.8% increase), was seen after cows infected with gastrointestinal nematodes, paramphistomes and Fasciola hepatica were treated with broad-spectrum anthelmintics. Three hundred and ninety pairs of cows
E E Guobadia et al.
Veterinary parasitology, 68(3), 269-282 (1997-02-01)
The 17kDa and 69kDa polypeptide antigens which are specific to Fasciola gigantica were excised from polyacrylamide gels and used for the immunization of rabbits to raise monospecific antisera against these polypeptides. These sera were labelled with horseradish peroxidase and used
Treatment of Dicrocoelium dendriticum with a combination of thiophanate and brotianide.
B Tinar et al.
The Veterinary record, 123(25), 650-651 (1988-12-17)
J V Mbuh et al.
Veterinary parasitology, 66(3-4), 171-179 (1996-11-15)
The profiles of antibody response and circulating antigen levels in goats infected with Fasciola gigantica were studied by enzyme-linked immunoelectrotransfer blot (EITB) and enzyme-linked immunosorbent assay (ELISA). In the antibody assay, sera from goats experimentally infected with F. gigantica were
C Paraud et al.
Veterinary journal (London, England : 1997), 180(2), 265-267 (2008-03-04)
Paramphistome infections are very common in ruminants and may induce clinical signs, but little is known about effective treatments. In this study, the efficacy of oxyclozanide against Calicophoron daubneyi was studied in goats and its activity tested against immature stages

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service