Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C12H2Br8O
CAS Number:
Molecular Weight:
801.38
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
208-759-1
MDL number:
grade
analytical standard
Quality Segment
shelf life
limited shelf life, expiry date on the label
concentration
50 μg/mL in isooctane
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
environmental
format
single component solution
storage temp.
2-8°C
SMILES string
Brc1cc(Oc2c(Br)c(Br)c(Br)c(Br)c2Br)cc(Br)c1Br
InChI
1S/C12H2Br8O/c13-4-1-3(2-5(14)6(4)15)21-12-10(19)8(17)7(16)9(18)11(12)20/h1-2H
InChI key
CVMKCYDBEYHNBM-UHFFFAOYSA-N
General description
BDE No 205 is a pollutant belonging to the class of polybrominated diphenyl ethers. It can be commercially used as a flame retardant additive in building materials, textiles, plastics and electronic equipment.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
BDE No 205 may be used as an analytical reference standard for the determination of the analyte in soil samples using high-performance liquid chromatography.
Other Notes
The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Central nervous system
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
10.4 °F
flash_point_c
-12 °C
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



