Skip to Content
Merck
All Photos(1)

Key Documents

33683

Supelco

BDE No 153 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,4,4′,5,5′-Hexabromodiphenyl ether solution, 2,2′,4,4′,5,5′-HexaBDE, PBDE 153

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Br6O
CAS Number:
Molecular Weight:
643.58
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1cc(Br)c(Oc2cc(Br)c(Br)cc2Br)cc1Br

InChI

1S/C12H4Br6O/c13-5-1-9(17)11(3-7(5)15)19-12-4-8(16)6(14)2-10(12)18/h1-4H

InChI key

RZXIRSKYBISPGF-UHFFFAOYSA-N

Related Categories

General description

BDE No 153 is a contaminant, which can also be commercially used as a flame retardant additive in building materials, textiles, plastics, and electronic equipment.

Application

BDE No 153 may be used as an analytical reference standard for the determination of the analyte in:
  • House dust samples using isotope dilution method combined with liquid chromatography coupled to negative ionization atmospheric pressure photoionization tandem mass spectrometry (LC-NI-APPI-MS/MS).
  • Adipose tissue samples using gas chromatography coupled to ion-trap mass spectrometry (GC-IT-MS/MS).

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F

Flash Point(C)

-12 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Development of a gas chromatography/ion trap mass spectrometry based method for the quantification of polybrominated diphenyl ethers and polychlorinated biphenyls in adipose tissue.
Naert C, et al.
Rapid Communications in Mass Spectrometry, 18(19), 2317-2332 (2004)
Isotope dilution method for determination of polybrominated diphenyl ethers using liquid chromatography coupled to negative ionization atmospheric pressure photoionization tandem mass spectrometry: validation and application to house dust.
Abdallah E-A M, et al.
Analytical Chemistry, 81(17), 7460-7469 (2009)
E Beristain-Montiel et al.
Journal of chromatography. A, 1477, 100-107 (2016-11-30)
New clean technologies are needed to determine concentration of organic pollutants without generating more pollution. A method to extract Persistent Organic Pollutants (POPs) from airborne particulate matter was developed using a novel technology recently patented called ultrasound assisted extraction micro-scale
Siqi Zhang et al.
Neurotoxicology and teratology, 73, 54-66 (2019-04-04)
Asexual freshwater planarians are an attractive invertebrate model for high-throughput neurotoxicity screening, because they possess multiple quantifiable behaviors to assess distinct neuronal functions. Planarians uniquely allow direct comparisons between developing and adult animals to distinguish developmentally selective effects from general
Youssef Oulhote et al.
Environment international, 119, 79-88 (2018-06-26)
Prenatal exposure to polybrominated diphenyl ethers (PBDEs) has been associated with cognitive deficits and behavioral problems in children. To date, no study has examined this exposure in association with neurobehavioral development in infants younger than 12 months assessed with observational tasks.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service