Skip to Content

33682

BDE No 126 solution

50 μg/mL in isooctane, analytical standard

Synonym(s):

3,3′,4,4′,5-Pentabromodiphenyl ether solution, 3,3′,4,4′,5-PentaBDE, PBDE 126

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H5Br5O
CAS Number:
Molecular Weight:
564.69
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
EC Number:
208-759-1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

50 μg/mL in isooctane

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Brc1ccc(Oc2cc(Br)c(Br)c(Br)c2)cc1Br

InChI

1S/C12H5Br5O/c13-8-2-1-6(3-9(8)14)18-7-4-10(15)12(17)11(16)5-7/h1-5H

InChI key

SJNIIWPIAVQNRK-UHFFFAOYSA-N

General description

BDE No 126 is widely used as a flame retardant additive in building materials, textiles, plastics, and electronic equipments.

Application

BDE No 126 may be used as an analytical reference standard for the determination of the analyte in environmental matrices using liquid chromatography coupled to negative ion spray ionization tandem mass spectrometry (LC-ISP(-)-MS/MS) with multiple reaction monitoring mode (MRM).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

The customer needs to make sure that the standard is homogeneous, this can be achieved by shaking, vortexing , or sonicating the ampoule before opening the ampoule. Care should be taken when opening the ampoule to avoid any injuries from the broken glass.
The standard should be transferred to a clean and appropriate vial or flask using clean pipettes or micro pipettes. The vial should be immediately capped to avoid any loss or evaporation of the solvent.
After opening the ampoule, the standard should not be stored or kept in the ampoule. To preserve the integrity of the product, the standard should be transferred to an appropriate vial that must be capped and stored according to the recommendation on the Certificate of Analysis.


signalword

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

10.4 °F

flash_point_c

-12 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library