Skip to Content
Merck
All Photos(1)

Documents

33491

Supelco

Dieldrin

PESTANAL®, analytical standard

Synonym(s):

1,2,3,4,10,10-Hexachloro-1,4,4a,5,6,7,8,8a-octahydro-6,7-epoxy-1,4:5,8-dimethanonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8Cl6O
CAS Number:
Molecular Weight:
380.91
Beilstein:
91396
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

13.2 (vs air)

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

143-144 °C (lit.)

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

[H][C@@]12O[C@]1([H])C3CC2[C@]4([H])[C@@]3([H])[C@]5(Cl)C(Cl)=C(Cl)[C@@]4(Cl)C5(Cl)Cl

InChI

1S/C12H8Cl6O/c13-8-9(14)11(16)5-3-1-2(6-7(3)19-6)4(5)10(8,15)12(11,17)18/h2-7H,1H2/t2-,3+,4+,5-,6-,7+,10+,11-

InChI key

DFBKLUNHFCTMDC-PICURKEMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dieldrin is a chlorinated hydrocarbon insecticide, which is one of the persistent contaminants present in the environment.

Application

Dieldrin may be used as a reference standard for the determination of the analyte in eggs using normal-phase high-performance liquid chromatography with photo-diode array detection (HPLC-PDA).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Normal-phase High-Performance Liquid Chromatographic determination and identification of Aldrin, Dieldrin, and DDTs in eggs.
Farusawa N.
Journal of Chromatographic Science, 39(5), 183-187 (2001)
Dieldrin: degradation by soil microorganisms.
Matsumura F and Boush MG
Science (New York, N.Y.), 156(3777), 959-961 (1967)
Toxics A to Z: A Guide to Everyday Pollution Hazards (1991)
Maria Teresa Baltazar et al.
Toxicology letters, 230(2), 85-103 (2014-02-08)
The etiology of most neurodegenerative disorders is multifactorial and consists of an interaction between environmental factors and genetic predisposition. The role of pesticide exposure in neurodegenerative disease has long been suspected, but the specific causative agents and the mechanisms underlying
Anumantha G Kanthasamy et al.
Neurotoxicology, 26(4), 701-719 (2005-08-23)
Parkinson's disease (PD) is increasingly recognized as a neurodegenerative disorder strongly associated with environmental chemical exposures. Recent epidemiological data demonstrate that environmental risk factors may play a dominant role as compared to genetic factors in the etiopathogenesis of idiopathic Parkinson's

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service