Skip to Content
Merck
All Photos(1)

Key Documents

33447

Supelco

Mecillinam

VETRANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O3S
CAS Number:
Molecular Weight:
325.43
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

storage temp.

2-8°C

SMILES string

[H][C@]12SC(C)(C)[C@@H](N1C(=O)[C@H]2\N=C\N3CCCCCC3)C(O)=O

InChI

1S/C15H23N3O3S/c1-15(2)11(14(20)21)18-12(19)10(13(18)22-15)16-9-17-7-5-3-4-6-8-17/h9-11,13H,3-8H2,1-2H3,(H,20,21)/b16-9+/t10-,11+,13-/m1/s1

InChI key

BWWVAEOLVKTZFQ-ISVUSNJMSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: ß-lactam
Mecillinam is a beta-lactam antibiotic, specific to penicillin-binding protein 2 (PBP2) in Escherichia coli, which tends to block cell wall elongation and indirectly blocks cell division.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Karen L Nielsen et al.
Journal of medical microbiology, 63(Pt 4), 582-589 (2014-01-28)
Urinary tract infections (UTIs) are primarily caused by Escherichia coli with the patient's own faecal flora acting as a reservoir for the infecting E. coli. Here we sought to characterize the E. coli faecal flora of UTI patients and healthy
Mecillinam resistance in Escherichia coli is conferred by loss of a second activity of the AroK protein
Vinella D, et al.
Journal of Bacteriology, 178, 3818-3828 (1996)
Christina A Ahlstrom et al.
Scientific reports, 8(1), 7361-7361 (2018-05-11)
Antimicrobial resistance (AMR) in bacterial pathogens threatens global health, though the spread of AMR bacteria and AMR genes between humans, animals, and the environment is still largely unknown. Here, we investigated the role of wild birds in the epidemiology of
Aurélie Scherler et al.
Microorganisms, 8(1) (2020-01-16)
Chlamydiae, such as Chlamydia trachomatis and Chlamydia pneumoniae, can cause chronic infections. It is believed that persistent forms called aberrant bodies (ABs) might be involved in this process. AB formation seems to be a common trait of all members of
Niki Lampri et al.
The Journal of antimicrobial chemotherapy, 67(10), 2424-2428 (2012-06-06)
Extended-spectrum β-lactamases (ESBLs) have emerged as an important mechanism of β-lactam resistance among community uropathogens. We characterized the ESBLs of a collection of Escherichia coli isolates recovered from outpatients with urinary tract infection during nationwide surveillance conducted from 2005 to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service