Skip to Content
Merck
All Photos(1)

Key Documents

33438

Supelco

Fusarenon X

analytical standard

Synonym(s):

3α,7α,15-Trihydroxy-4β-acetoxy-12,13-epoxytrichothec-9-en-8-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22O8
CAS Number:
Molecular Weight:
354.35
MDL number:
UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC(=O)O[C@@H]1[C@@H](O)[C@H]2O[C@@H]3C=C(C)C(=O)[C@@H](O)[C@]3(CO)[C@]1(C)[C@]24CO4

InChI

1S/C17H22O8/c1-7-4-9-16(5-18,12(22)10(7)20)15(3)13(24-8(2)19)11(21)14(25-9)17(15)6-23-17/h4,9,11-14,18,21-22H,5-6H2,1-3H3/t9?,11-,12-,13-,14?,15?,16?,17?/m1/s1

InChI key

XGCUCFKWVIWWNW-LTNYHXKASA-N

General description

Fusarenon X is a trichothecene mycotoxin isolated from Fusarium nivale.

Application

Fusarenon X may be used as a reference standard for the determination of fusarenon X in barley, wheat and corn samples by high-performance thin-layer chromatography (HPTLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-performance thin-layer chromatographic determination of deoxynivalenol, fusarenon-X, and nivalenol in barley, corn, and wheat.
Trucksess M W, et al.
Journal of Agricultural and Food Chemistry, 35(4), 445-448 (1987)
General pharmacological studies of fusarenon-X, a trichothecene mycotoxin from Fusarium species.
Matsuoka Y, et al.
Toxicology and Applied Pharmacology, 50(1), 87-94 (1979)
N Tani et al.
Microbiology and immunology, 39(8), 635-637 (1995-01-01)
The effect of trichothecene mycotoxins, deoxynivalenol (DON), fusarenon-X (FX) and nivalenol (NIV), on plaque formation of herpes simplex virus types 1 and 2 (HSV-1 and HSV-2) in HEp-2 cells was examined. The 50% effective concentrations (EC50) of DON, FX, and
J Li et al.
The Journal of veterinary medical science, 59(3), 191-199 (1997-03-01)
Male 5-week-old Wistar rats orally (po) administered with fusarenon-X (FX) 1.5 mg/kg and control rats po-treated with distilled water were sacrificed at 0-48 hr after gavage. FX-administered rats showed significant dilatation of the stomach with increased fluid contents at 1-24
Margit Schollenberger et al.
International journal of food microbiology, 72(1-2), 85-89 (2002-02-15)
A total of 60 samples of wheat flour were collected during the first 6 months of 1999 from mills and food stores in an area in southwest Germany. Samples included whole-grain and two types of white flour with these three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service