Skip to Content
Merck
All Photos(1)

Key Documents

32964

Supelco

Metrafenone

PESTANAL®, analytical standard

Synonym(s):

(3-Bromo-6-methoxy-2-methylphenyl)(2,3,4-trimethoxy-6-methylphenyl)methanone, 3′-Bromo-2,3,4,6′-tetramethoxy-2′,6-dimethylbenzophenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H21BrO5
CAS Number:
Molecular Weight:
409.27
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccc(Br)c(C)c1C(=O)c2c(C)cc(OC)c(OC)c2OC

InChI

1S/C19H21BrO5/c1-10-9-14(23-4)18(24-5)19(25-6)15(10)17(21)16-11(2)12(20)7-8-13(16)22-3/h7-9H,1-6H3

InChI key

AMSPWOYQQAWRRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metrafenone is a novel benzophenone fungicide that could be used against powdery mildew and eye spot in vegetables and fruits.

Application

Metrafenone may be used as an analytical reference standard for the determination of the analyte in:
  • Vegetables by gas chromatography coupled to electron capture detector (GC-ECD).
  • Honeybees by liquid and gas chromatography combined with tandem mass spectrometry (LC-MS/MS and GC-MS/MS).
  • Animal samples and earthworms by LC-MS/MS.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A liquid chromatography-electrospray tandem mass spectrometry method for the determination of multiple pesticide residues involved in suspected poisoning of non-target vertebrate wildlife, livestock and pets.
Taylor MJ, et al.
Analytical Methods : Advancing Methods and Applications, 5(1), 248-259 (2013)
Anna Markheiser et al.
Insects, 11(1) (2020-01-16)
The identification of volatile organic compounds (VOCs) leading to short-range attraction and oviposition of the European grapevine moth Lobesia botrana and European grape berry moth Eupoecilia ambiguella (Lepidoptera: Tortricidae) is crucial in order to establish bait-based decision support systems for
Multi-residue method for the determination of pesticides and pesticide metabolites in honeybees by liquid and gas chromatography coupled with tandem mass spectrometry?Honeybee poisoning incidents.
Kiljanek T, et al.
Journal of Chromatography A, 1435(1), 100-114 (2016)
Development of a method for the simultaneous determination of multi-class pesticides in earthworms by liquid chromatography coupled to tandem electrospray mass spectrometry.
Daniele G, et al.
Analytical and Bioanalytical Chemistry, 410(20), 5009-5018 (2018)
Determination of residual levels of metrafenone in lettuce grown under greenhouse conditions using gas chromatography with a micro-electron capture detector.
Kabir MH, et al.
Journal of Applied Biological Chemistry, 59(1), 43-49 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service