Skip to Content
Merck
All Photos(1)

Key Documents

32542

Supelco

Menidazole

VETRANAL®, analytical standard

Synonym(s):

2-Methyl-4(5)-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H5N3O2
CAS Number:
Molecular Weight:
127.10
Beilstein:
4032
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cc1ncc([nH]1)[N+]([O-])=O

InChI

1S/C4H5N3O2/c1-3-5-2-4(6-3)7(8)9/h2H,1H3,(H,5,6)

InChI key

FFYTTYVSDVWNMY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daniel D Rossouw et al.
Nuclear medicine and biology, 32(4), 385-394 (2005-05-10)
The inflammation- and infection-seeking properties of (131)I-labeled ornidazole, a 5-nitroimidazole derivative, have recently been reported. Whole-body images in rabbits showed a more rapid uptake in inflamed areas compared to (67)Ga. In the present study, two novel (123)I-labeled 2-methyl-4-nitroimidazole derivatives were
Fa-qing Ye et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 38(4), 260-263 (2003-08-02)
To study the synthesis and antibacterial activity of pyridonecarboxylic acid derivatives containing 2-methyl-5-nitroimidazol. Pyridonecarboxylic acid derivatives containing 2-methyl-5-nitroimidazol were synthesized primarily from 2-methyl-5-nitroimidazol, norfloxacin, ciprofloxacin, enoxacin via nucleophilic substitution and esterification. The antibacterial activity of the nine target compounds were
G Carignan et al.
Food additives and contaminants, 8(4), 467-475 (1991-07-01)
An unidentified metabolite of dimetridazole (DMZ), found in pig plasma, muscle and kidney, was shown by chromatography and spectroscopy to be 2-methyl-5-nitroimidazole (2-MNI), resulting from N-demethylation of DMZ. This route of degradation competes with the oxidation pathway previously described. The
Fa-qing Ye et al.
Yao xue xue bao = Acta pharmaceutica Sinica, 40(2), 132-135 (2005-05-07)
To study the synthesis and antibacterial activity of ciprofloxacin derivatives. Ciprofloxacin derivatives were synthesized primarily from 2-methyl-5-nitroimidazol and ciprofloxacin through nucleophilic substitution. The antibacterial activity of the synthesized compounds were tested. Nine new compounds were synthesized. The structure of the
Madhava B Mallia et al.
Bioorganic & medicinal chemistry, 14(23), 7666-7670 (2006-09-05)
An iminodiacetic acid (IDA) derivative of 2-methyl-5-nitroimidazole was synthesized as a carrier molecule for radiolabeling with the gamma emitting radioisotope, 99mTc, for imaging hypoxic regions of tumors. The ligand was synthesized in excellent yield and labeled using freshly prepared [99mTc(CO)3(H2O)3]+

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service