Skip to Content
Merck

32173

Ipronidazole

VETRANAL®, analytical standard

Synonym(s):

1-Methyl-2-(1-methylethyl)-5-nitro-1H-imidazole, 2-Isopropyl-1-methyl-5-nitroimidazol, NSC 109212, Ro 07-1554

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H11N3O2
CAS Number:
Molecular Weight:
169.18
PubChem Substance ID:
UNSPSC Code:
41116107
NACRES:
NA.24
EC Number:
238-957-3
MDL number:
Beilstein/REAXYS Number:
744577
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

technique(s)

HPLC: suitable, chromatography: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

[N+](=O)([O-])c1[n](c(nc1)C(C)C)C

InChI

1S/C7H11N3O2/c1-5(2)7-8-4-6(9(7)3)10(11)12/h4-5H,1-3H3

InChI key

NTAFJUSDNOSFFY-UHFFFAOYSA-N

General description

Ipronidazole is a veterinary drug, which is widely used in controlling histomoniasis in turkeys.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Ipronidazole may be used as a reference standard in the determination of ipronidazole in tissue samples using high-performance liquid chromatography coupled with an ultraviolet detector (HPLC) and thermospray tandem mass spectrometry (HPLC-MS/MS).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library