Skip to Content
Merck
All Photos(3)

Key Documents

319945

Sigma-Aldrich

trans-1,2-Diaminocyclohexane-N,N,N′,N′-tetraacetic acid monohydrate

ACS reagent, for complexometry, 98%

Synonym(s):

1,2-Cyclohexanedinitrilotetraacetic acid, Chel-CD, DCTA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H22N2O8 · H2O
CAS Number:
Molecular Weight:
364.35
Beilstein:
3222748
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent
for complexometry

Quality Level

Assay

97.5-100.5%
98%

form

powder

ign. residue

≤0.2%

mp

213-216 °C (lit.)

cation traces

Fe: ≤0.005%
heavy metals (as Pb): ≤0.001%

functional group

amine
carboxylic acid

SMILES string

[H]O[H].OC(=O)CN(CC(O)=O)[C@@H]1CCCC[C@H]1N(CC(O)=O)CC(O)=O

InChI

1S/C14H22N2O8.H2O/c17-11(18)5-15(6-12(19)20)9-3-1-2-4-10(9)16(7-13(21)22)8-14(23)24;/h9-10H,1-8H2,(H,17,18)(H,19,20)(H,21,22)(H,23,24);1H2/t9-,10-;/m1./s1

InChI key

VASZYFIKPKYGNC-DHTOPLTISA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1,2-Diaminocyclohexane-N,N,N,N′-tetraacetic acid monohydrate is a multidentate ligand{1-6} that may be used as a chelating agent in the preparation of metal-chelate complexes.
Ligand used to prepare lanthanide shift reagents.

Legal Information

Chel is a trademark of Ciba-Geigy AG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of Lanthanide Shift Reagents with 31P FT-NMR Spectroscopy To Analyze Concentrated Lake-Water Samples.
M A Nanny et al.
Environmental science & technology, 28(8), 1521-1527 (1994-08-01)
Monoclonal antibodies that recognize minimal differences in the three-dimensional structures of metal? chelate complexes.
Jones RM, et al.
Bioconjugate Chemistry, 13(3), 408-415 (2002)
Permeation Liquid Membrane Metal Transport: Studies of Complex Stoichiometries and Reactions in Cu (II) Extraction with the Mixture 22DD? Laurate in Toluene/Phenylhexane.
Guyon F, et al.
Analytical Chemistry, 72(6), 1328-1333 (2000)
Protonation constants of 1-hydroxyethylidene-1, 1-diphosphonic acid, diethylenetriamino-N, N, N?, N ?, N ?-penta-acetic trans-1, 2-diaminocyclohexane-N, N, N?, N?-tetra-acetic acid.
Mioduski T.
Talanta, 27(3), 299-303 (1980)
Touraj Aligholipour Farzani et al.
STAR protocols, 1(3), 100209-100209 (2020-12-31)
We describe the production of single-cycle (sc) and replication-competent recombinant vesicular stomatitis viruses (rcVSVs) displaying heterologous envelope glycoproteins (Envs) on their surface. We prepare scVSVs by transiently expressing HIV-1 Envs or SARS-CoV-2 spike followed by infection of the cells with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service