Skip to Content
Merck
All Photos(1)

Key Documents

31510

Supelco

Bupirimate

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H24N4O3S
CAS Number:
Molecular Weight:
316.42
Beilstein:
758056
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCCCc1c(C)nc(NCC)nc1OS(=O)(=O)N(C)C

InChI

1S/C13H24N4O3S/c1-6-8-9-11-10(3)15-13(14-7-2)16-12(11)20-21(18,19)17(4)5/h6-9H2,1-5H3,(H,14,15,16)

InChI key

DSKJPMWIHSOYEA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Bupirimate is an active ingredient in plant protection products and is commonly used as a systemic fungicide. It belongs to the family of pyrimidines and finds applications in controlling powdery mildew in roses and apple plantations.

Application

Bupirimate may be used as a reference standard in the determination of bupirimate in fruits using gas chromatography coupled with a flame photometric detector (GC-FPD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Aquatic Chronic 1 - Carc. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

chromatography/quadrupole-time-of-flight mass spectrometry},
Ernst.FG, et al.
Journal of Chromatography A, 179(2), 351-354 (1979)
Detoxification of bupirimate pesticide in aqueous solutions by electrochemical oxidation
Errami.M, et al.
Der Pharm Chem, 4, 297-310 (2012)
Yun-Hui Teng et al.
Chang Gung medical journal, 27(12), 877-886 (2005-03-10)
The purpose of this retrospective study was to determine whether epidural fentanyl-bupivacaine patient-controlled analgesia (PCA) was more efficacious and had fewer adverse effects than epidural or intravenous morphine PCA. We retrospectively retrieved data from 859 patients (mean age 64+/-7 years)
Bupirimate--a new fungicide for the control of powdery mildews on apples and other crops
R.JF, et al.
Proceedings of the British Insecticide and Fungicide Conference (1975)
L J Fitzpatrick et al.
Journal of chromatography. A, 918(2), 429-433 (2001-06-16)
This paper assesses the effect of pressurised fluid extraction (PFE) on the recovery of bupirimate and its degradation product, ethirimol from a range of soil types. The analytes were extracted under standard conditions (pressure, 2000 p.s.i.; temperature, 100 degrees C;

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service