Skip to Content
Merck
All Photos(1)

Key Documents

31430

Supelco

Dextran

analytical standard, for GPC, Set Mp 1,000-400,000

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[C6H10O5]n
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard
for GPC

Quality Level

mol wt

Mp 1,000-400,000

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

food and beverages

format

neat

SMILES string

O1C(C(C(C(C1CO)O)O)O)OCC2OC(C(C(C2O)O)O)OCC(O)C(O)C(O)C(O)C=O

InChI

1S/C18H32O16/c19-1-5(21)9(23)10(24)6(22)3-31-17-16(30)14(28)12(26)8(34-17)4-32-18-15(29)13(27)11(25)7(2-20)33-18/h1,5-18,20-30H,2-4H2

InChI key

FZWBNHMXJMCXLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dextran is an exopolysaccharide synthesized by lactic acid bacteria from sucrose. It consists of repeating units of D-glucose with the monomers joined to each other through α-1,6 linkages. In addition, the linear D-glucose chains show branching at frequent intervals, linked by α-1,4, α-1,3, or α-1,2 linkages.

Application

The analytical standards can also be used as follows:
  • High-performance size exclusion chromatography (HPSEC) based determination of the molecular weight of a dextran synthesized by Leuconostoc citreum B-2, isolated from a fermentation product of pineapple
  • Determination of molecular weight of polysaccharides obtained from four Auricularia species by high-performance liquid chromatography using a water-soluble gel column
  • Identification of molecular weight of a water-soluble polysaccharide isolated by anion exchange chromatography from the dried leaves of C. paliurus by gel permeation chromatography (GPC)
  • Gel permeation chromatography (GPC) based molecular weight measurement of a dextran produced by two strains of Leuconostoc mesenteroides
  • Estimation of molecular weight of a water-soluble polysaccharide obtained from thinned-young apple by high-performance size exclusion chromatography (HPSEC)

Components

set contains 10 different dextran standards with 500 mg with Mp ~1,100 (Cat. No. 31416), ~4,400 (Cat. No. 31417), ~10,000 (Cat. No. 31418), ~20,000 (Cat. No. 31419), ~45,000 (Cat. No. 31420),~65,000 (Cat. No. 31421), ~125,000 (Cat. No. 31422), ~195,000 (Cat. No. 31423), ~275,000 (Cat. No. 31424), ~400,000 (Cat. No. 31425); The components can also be ordered separately under the product numbers indicated.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Yu et al.
Molecular human reproduction, 20(3), 260-270 (2013-11-26)
Accumulating evidence indicates that reduced fecundity associated with endometriosis reflects a failure of embryonic receptivity. Microdomains composed of endometrial gap junctions, which facilitate cell-cell communication, may be implicated. Pharmacological or genetic inhibition of connexin (Cx) 43 block human endometrial cell
Erik C B Johnson et al.
Molecular neurodegeneration, 15(1), 53-53 (2020-09-15)
Alzheimer's disease (AD) is the most frequent and costly neurodegenerative disorder. Although diverse lines of evidence suggest that the amyloid precursor protein (APP) is involved in its causation, the precise mechanisms remain unknown and no treatments are available to prevent
Fangwei Gu et al.
The Plant cell, 28(7), 1722-1737 (2016-06-30)
In plants, the presence of a load-bearing cell wall presents unique challenges during cell division. Unlike other eukaryotes, which undergo contractile cytokinesis upon completion of mitosis, plants instead synthesize and assemble a new dividing cell wall to separate newly formed
Mingshu Sun et al.
International journal of immunopathology and pharmacology, 30(3), 227-237 (2017-01-31)
Systemic inflammation is involved in vascular calcification and cardiovascular disease which is the leading cause of mortality in rheumatoid arthritis (RA). A high level of serum interleukin (IL)-6 plays a key role in local and systemic inflammation in RA. However
Yoan Arribat et al.
The Journal of clinical investigation, 129(12), 5312-5326 (2019-09-11)
Growing evidence shows that alterations occurring at early developmental stages contribute to symptoms manifested in adulthood in the setting of neurodegenerative diseases. Here, we studied the molecular mechanisms causing giant axonal neuropathy (GAN), a severe neurodegenerative disease due to loss-of-function

Related Content

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Dextrans are polysaccharides with molecular weights ≥1,000 Dalton, featuring a linear backbone of α-linked d-glucopyranosyl repeating units.

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service