Skip to Content
Merck
All Photos(2)

Documents

30552

Supelco

Monensin methyl ester

Selectophore, ≥97.0% (TLC)

Synonym(s):

Methyl monensin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C37H64O11
CAS Number:
Molecular Weight:
684.90
Beilstein:
3641733
UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

Assay

≥97.0% (TLC)

storage temp.

2-8°C

SMILES string

CC[C@]1(CCC(O1)[C@]2(C)CC[C@]3(C[C@H](O)[C@@H](C)C(O3)[C@@H](C)[C@@H](OC)[C@H](C)C(=O)OC)O2)C4OC(C[C@@H]4C)C5O[C@@](O)(CO)[C@H](C)C[C@@H]5C

InChI

1S/C37H64O11/c1-11-35(32-21(3)17-27(44-32)29-20(2)16-22(4)37(41,19-38)47-29)13-12-28(45-35)34(8)14-15-36(48-34)18-26(39)23(5)31(46-36)24(6)30(42-9)25(7)33(40)43-10/h20-32,38-39,41H,11-19H2,1-10H3/t20-,21+,22+,23+,24+,25+,26-,27-,28+,29-,30-,31-,32-,34+,35-,36+,37-/m0/s1

InChI key

PFRZSHIENRKVSE-RJTHVKINSA-N

General description

Monensin, is an antibiotic produced by Streptomyces cinnamonensis. It is a naturally occurring ionophore. It has an acyclic polyether sequence and a terminal carboxylic acid moiety. It has the ability to effectively carry protons and cations through the cell membrane, hence it adequately harbours Na+ into a characteristic pseudoactivity, thereby effectively transporting it across a bio-membrane. It also forms stable complexes with k+, Rb+ and Ag+; hence it can be chemically modified to form cation-binding sites, thereby attaining transport functionalities.
Visit our Sensor Applications portal to learn more.

Application


  • The selectivity of membrane ion-selective electrodes: This research explores the use of temperature variations to adjust the selectivity of ion-selective electrodes, utilizing Monensin methyl ester as a key component for sodium ion detection (Zahran et al., 2010).

  • Spectroscopic and semiempirical studies of a proton channel formed by the methyl ester of monensin A.: This paper presents a detailed analysis of the proton channel properties of Monensin methyl ester through spectroscopic and computational methods, highlighting its potential in analytical chemistry applications (Huczyński et al., 2006).

  • Ion chromatography detector based on solid-state ion-selective electrode array.: The development of an ion chromatography detector employing solid-state ion-selective electrodes, with Monensin methyl ester playing a crucial role in sodium ion detection, is detailed in this study (Lee et al., 2000).

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adam Huczyñski et al.
The journal of physical chemistry. B, 110(31), 15615-15623 (2006-08-04)
Monensin A is an ionophore able to carry protons and cations through the cell membrane. Its methyl ester (MON1) and its hydrates have been studied in acetonitrile, and its deuterated analogue by Fourier transform infrared (FTIR) and (1)H and (13)C
José N Sousa et al.
European journal of mass spectrometry (Chichester, England), 13(3), 191-198 (2007-09-21)
This paper reports the first comparative study of the gas-phase fragmentation chemistry of monensin in negative ion mode electrospray and nanoelectrospray tandem mass spectrometry. The fragmentation was observed to occur at lower energies in nanoelectrospray than electrospray. The major product
Biosynthesis of monensin.
Day LE
Antimicrobial Agents and Chemotherapy, 4 (4), 410-414 (1973)
Specific Ag+ ion-transport properties of functionalized biological monensin ionophore.
Tsukube H and Sohmiya H.
Inorgorganica Chimica Acta, 166 (1), 27-29 (1989)
Spectroscopic and semiempirical studies of a proton channel formed by the methyl ester of monensin A.
Huczynski A
The Journal of Physical Chemistry B, 110 (31), 15615-15623 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service