24901
Chloride ionophore II
Selectophore™, function tested
Synonym(s):
4,5-Dimethyl-3,6-dioctyloxy-o-phenylene-bis(mercurytrifluoroacetate), ETH 9009
About This Item
Recommended Products
grade
for ion-selective electrodes
Quality Level
product line
Selectophore™
form
powder
quality
function tested
mp
177-179 °C
SMILES string
CCCCCCCCOc1c(C)c(C)c(OCCCCCCCC)c([Hg]OC(=O)C(F)(F)F)c1[Hg]OC(=O)C(F)(F)F
InChI
1S/C24H40O2.2C2HF3O2.2Hg/c1-5-7-9-11-13-15-19-25-23-17-18-24(22(4)21(23)3)26-20-16-14-12-10-8-6-2;2*3-2(4,5)1(6)7;;/h5-16,19-20H2,1-4H3;2*(H,6,7);;/q;;;2*+1/p-2
InChI key
UMYVIFNPTFWRLT-UHFFFAOYSA-L
General description
Application
Packaging
Other Notes
Legal Information
Disclaimer

Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2
Storage Class Code
6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service