Skip to Content
Merck
All Photos(1)

Key Documents

217417

Supelco

AmberChrom 1X8 Ion Exchange Resin

chloride form, 50-100 mesh

Synonym(s):

AmberChrom 1X8 chloride form, AmberChrom 1X8-100 ion exchange resin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C10H12 · C10H10 · C8H8 · C3H9N)X
CAS Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

AmberChrom 1X8 50-100 MESH (CL-) Anion,

description

cross-linkage 8%

Quality Level

form

beads

crosslinking

8 % cross-linked

parameter

66 °C OH- form max. temp.

moisture

43-48%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

50-100 mesh

operating pH

0-14

capacity

1.2 meq/mL by wetted bed volume

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

AmberChrom 1X8 is a strongly basic anion exchange resin which is used as an absorbent in solid phase extraction.

Application

AmberChrom 1X8 chloride form was used:
  • to separate the 3H2O from D[2-3H] glucose using Dowex columns
  • in a assay to determine the pantothenate kinase using Dowex column.
  • to separate inositol phosphates (IPs) from myo-[3H]inositol by Dowex chromatography.
Strongly basic (type I) anion exchange resin for fine chemical processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

strongly basic anion.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L M King et al.
Cardiovascular research, 39(2), 381-392 (1998-11-03)
Experimental data from isolated rat hearts suggest that glycolysis in severe myocardial ischemia is inhibited by accumulation of glycolytic metabolites. In contrast, positron emission tomography (PET) in patients with myocardial ischemia records a 'mismatch' between the decreased coronary flow in
M N Fisher et al.
FEBS letters, 190(2), 293-296 (1985-10-14)
The relative tissue activities of pantothenate kinase range from 1 to 5 nmol . min-1 . g-1 wet wt for heart, brain, kidney, and liver. The enzyme partially purified from each tissue is inhibited by CoA, but there is a
A Cedazo-Mínguez et al.
FEBS letters, 504(1-2), 45-49 (2001-08-28)
The mechanism(s) by which the E4 isoform of apolipoprotein E (apoE4) influences Alzheimer's disease (AD) are not fully known. We report that apoE4, but not apoE3, disrupts carbachol-stimulated phosphoinositide (PI) hydrolysis in SH-SY5Y neuroblastoma cells. Carbachol responses were also disrupted
Ayşem Uzer et al.
Forensic science international, 174(2-3), 239-243 (2007-06-09)
The Meisenheimer anions formed from TNT in KOH solutions in alcohol or acetone were used in screening tests for TNT among possible nitro-explosives. The same reaction was used for the spectrophotometric assay of TNT in soil by CRREL (Cold Regions

Related Content

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Low pressure liquid chromatography (LPLC) operates at low pressures, using a pump to drive the mobile phase onto the column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service