Skip to Content
Merck

217395

AmberChrom 1X2 Ion Exchange Resin

chloride form, 200-400 mesh

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 1X2 chloride form, 200-400 mesh

form

beads

Quality Segment

crosslinking

2 % cross-linked

parameter

66 °C OH- form max. temp., 99 °C Cl- form max. temp.

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

trimethylbenzylammonium functional group

particle size

200-400 mesh

operating pH

0-14

capacity

0.7 meq/mL by wetted bed volume, 65-75 % water retention capacity

separation technique

anion exchange

SMILES string

N(C)(C)C.C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8.C3H9N/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8;1-4(2)3/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2;1-3H3

InChI key

HADXLRSCRPYPJJ-UHFFFAOYSA-N

General description

AmberChrom 1X2 is an anion exchange resin.

Application

AmberChrom 1X2 chloride form was used to separate cadmium from seawater matrix using column chromatography. It was used in column for gas chromatography-mass spectrometric analysis.
Strongly basic (type I) anion exchange resin for food & drug processing; meets requirements of FDA Food Additive Regulation 21 CFR 173.25

Features and Benefits

strongly basic anion.

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberchrom is a trademark of DuPont de Nemours, Inc.


Still not finding the right product?

Explore all of our products under AmberChrom 1X2 chloride form


Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library