Skip to Content
Merck
All Photos(1)

Documents

21719

Supelco

Methyl octanoate

analytical standard

Synonym(s):

Caprylic acid methyl ester, Methyl caprylate, Octanoic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6COOCH3
CAS Number:
Molecular Weight:
158.24
Beilstein:
1752270
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.8% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.418

bp

193-194 °C (lit.)

density

0.877 g/mL at 20 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

CCCCCCCC(=O)OC

InChI

1S/C9H18O2/c1-3-4-5-6-7-8-9(10)11-2/h3-8H2,1-2H3

InChI key

JGHZJRVDZXSNKQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl octanoate, also known as methyl caprylate, is an octanoate ester and belongs to the group of fatty acid methyl esters (FAMEs). It is naturally found in Saccharomyces cerevisiae and other organisms.

Application

This analytical standard can also be used as follows:
  • Gas chromatography-tandem differential mobility spectrometry (DMS) based separation and quantification of 16 methyl- and ethyl- fatty acid esters from biodiesel samples
  • Development and validation of a gas chromatography-flame ionization (GC-FID) method to determine fatty acid methyl esters (FAMEs) in beer wort samples following their solid phase extraction and BF3 based methylation
  • Comparative analysis of gas chromatography-combustion-mass spectrometry and gas chromatography-flame ionization detection methods for the determination of fatty acid methyl esters (FAMEs) in biodiesel samples
  • Identification and quantification of fatty acids in walnut and coconut oil samples by gas chromatography-mass spectrometry (GC-MS) after their methyl esterification by BF3-MeOH method
  • Simultaneous determination of fatty acids in bovine colostrum samples by GC-FID after their derivatization to ester forms using an acidic catalyst boron trifluoride

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Elliott et al.
Biochimica et biophysica acta, 978(2), 337-340 (1989-01-30)
The effects of n-alkanes (n-pentane to n-octane), n-alkanols (n-pentanol to n-undecanol) and two carboxylic esters (methyl pentanoate and methyl octanoate) on the conductance of squid giant axons in a high potassium, zero sodium bathing solution have been examined. Sodium and
C Togbé et al.
The journal of physical chemistry. A, 114(11), 3896-3908 (2010-03-20)
There is a growing interest for using bioethanol-biodiesel fuel blends in diesel engines but no kinetic data and model for their combustion were available. Therefore, the kinetics of oxidation of a biodiesel-bioethanol surrogate fuel (methyl octanoate-ethanol) was studied experimentally in
T J Gianfagna et al.
Life support & biosphere science : international journal of earth space, 5(2), 255-261 (2001-09-07)
Given the constraints on physical space, energy supply, and labor availability, crop production efficiency must be maximized if a sustainable supply of food is to be produced on lunar and planetary space stations. In the modules designed for plant growth
D A Haydon et al.
The Journal of physiology, 341, 411-427 (1983-08-01)
The effects of several n-alkanols and n-alkyl oxyethylene alcohols, methyl octanoate, glycerol 1-monooctanoate and dioctanoyl phosphatidylcholine on the ionic currents and electrical capacity of the squid giant axon membrane have been examined. The peak inward current in voltage-clamped axons was
Oier Aizpurua-Olaizola et al.
Journal of food science, 80(1), E101-E107 (2014-12-05)
In this study, supercritical fluid extraction has been successfully applied to a sequential fractionation of fatty acids and polyphenols from wine wastes (2 different vitis vinifera grapes). To this aim, in a 1st step just fatty acids were extracted and

Protocols

HPLC Analysis of Fatty Acid Methyl Esters (FAMES) on SUPELCOSIL™ LC-18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service