Skip to Content
Merck

2106

Heparin sodium salt from porcine intestinal mucosa

endotoxin, free

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352201
NACRES:
NA.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Porcine intestinal mucosa

Quality Segment

form

powder

usage

sufficient for 5 mL blood anticoagulant

packaging

preweighed vial of 300 USP units

impurities

endotoxin, free

color

beige

solubility

water: soluble 50 g/L, acetone: insoluble, alcohol: insoluble, benzene: insoluble, chloroform: insoluble, diethyl ether: insoluble

compatibility

for use with E-Toxate™

storage temp.

room temp

SMILES string

[Na+].[S](=O)(=O)(NC1C(OC(C(C1O[S](=O)(=O)O)OC4OC(C(C(C4O[S](=O)(=O)O)O)O)C(=O)O)CO)OC2C(OC(C(C2O)O[S](=O)(=O)O)OC3C(OC(C(C3O)NC(=O)C)O)CO[S](=O)(=O)O)C(=O)O)O

InChI

1S/C26H42N2O37S5.Na/c1-4(30)27-7-9(31)13(6(56-23(7)39)3-55-67(43,44)45)58-26-19(65-70(52,53)54)12(34)16(20(62-26)22(37)38)60-24-8(28-66(40,41)42)15(63-68(46,47)48)14(5(2-29)57-24)59-25-18(64-69(49,50)51)11(33)10(32)17(61-25)21(35)36;/h5-20,23-26,28-29,31-

InChI key

JRTRSJGZMRQDHI-UHFFFAOYSA-N

Application

Heparin sodium salt from porcine intestinal mucosa has been used as a M199 medium supplement used for maintaining human umbilical vein endothelial cells.

Biochem/physiol Actions

Heparin sodium salt is the salt form of heparinic acid and is a polymer classified as a mucopolysaccharide or a glycosoaminoglycan. It is an anticoagulant that produces its major anticoagulant effect by activating antithrombin. Heparin binds to antithrombin III, a naturally occurring plasma protease inhibitor and accelerates significantly the rate at which antithrombin III (AT-III) inhibits coagulation proteases (factor Xa and thrombin). Additionally, it also facilitates the stabilization and regulation of tryptase as an enzymatically active tetramer.

Other Notes

To gain a comprehensive understanding of our extensive range of Polysaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Legal Information

E-Toxate is a trademark of Sigma-Aldrich Co. LLC

Disclaimer

Not for injection.


Still not finding the right product?


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library