Skip to Content
Merck

201871

Direct yellow 27

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H20N4Na2O9S3
CAS Number:
Molecular Weight:
662.62
PubChem Substance ID:
UNSPSC Code:
12171500
Colour Index Number:
13950
NACRES:
NA.47
EC Number:
233-461-3
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

λmax

393 nm

ε (extinction coefficient)

53000-57000 at 224-230 nm in water at 0.02 g/L

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].COc1ccccc1NC(=O)C(\N=N\c2ccc(cc2S([O-])(=O)=O)-c3nc4ccc(C)c(c4s3)S([O-])(=O)=O)=C(/C)O

InChI

1S/C25H22N4O9S3.2Na/c1-13-8-10-18-22(23(13)41(35,36)37)39-25(27-18)15-9-11-17(20(12-15)40(32,33)34)28-29-21(14(2)30)24(31)26-16-6-4-5-7-19(16)38-3;;/h4-12,30H,1-3H3,(H,26,31)(H,32,33,34)(H,35,36,37);;/q;2*+1/p-2/b21-14-,29-28+;;

InChI key

VHKQQPDOJAIOKS-NOBNIHJLSA-L

General description

As a direct dye, Direct Yellow 27 is an acid dye with large molecules that will bind to cellulose fibers directly. It is mono azo dye with good light fastness.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library