Skip to Content
Merck
All Photos(1)

Documents

18079

Millipore

Cycloheximide solution

0.1%, suitable for microbiology

Synonym(s):

CHX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23NO4
CAS Number:
Molecular Weight:
281.35
MDL number:
UNSPSC Code:
41171614
PubChem Substance ID:
NACRES:
NA.85

sterility

sterile (Filtered and Aseptic Handled)

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

concentration

0.1%

antibiotic activity spectrum

fungi

application(s)

environmental
food and beverages

microbiology

storage temp.

2-8°C

suitability

bacteria

SMILES string

O=C1[C@@]([C@H](O)CC2CC(NC(C2)=O)=O)([H])C[C@@H](C)C[C@@H]1C

InChI

1S/C15H23NO4/c1-8-3-9(2)15(20)11(4-8)12(17)5-10-6-13(18)16-14(19)7-10/h8-12,17H,3-7H2,1-2H3,(H,16,18,19)/t8-,9-,11-,12+/m0/s1

InChI key

YPHMISFOHDHNIV-FSZOTQKASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Cycloheximide solution is a selective supplement, that is safer and easier to use than cycloheximide powder. The required volume can be simply added to the medium before or after sterilization.

Application

Cycloheximide solution has been used as a supplement in De Man, Rogosa and Sharpe (MRS) and GM-17 agar plates to study its effects on lactic acid bacteria (LAB).

Biochem/physiol Actions

Cycloheximide solution is used to block the protein synthesis for suppressing the growth of fungi.

Physical form

Sterile filtered solution containing 0.1% cycloheximide

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V O Kaminskyy et al.
Cell death & disease, 4, e522-e522 (2013-03-09)
TNF-related apoptosis-inducing ligand (TRAIL) is a promising cytokine for killing tumor cells. However, a number of studies have demonstrated that different cancer cells resist TRAIL treatment and, moreover, TRAIL can promote invasion and metastasis in resistant cells. Here we report
Xing Wang et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(43), 15438-15443 (2014-10-16)
The receptor-interacting kinase-3 (RIP3) and its downstream substrate mixed lineage kinase domain-like protein (MLKL) have emerged as the key cellular components in programmed necrotic cell death. Receptors for the cytokines of tumor necrosis factor (TNF) family and Toll-like receptors (TLR)
Taichi Hara et al.
Scientific reports, 4, 6992-6992 (2014-11-12)
Peripheral myelin protein 22 (PMP22) resides in the plasma membrane and is required for myelin formation in the peripheral nervous system. Many PMP22 mutants accumulate in excess in the endoplasmic reticulum (ER) and lead to the inherited neuropathies of Charcot-Marie-Tooth
Julien Dubrulle et al.
eLife, 4 (2015-04-15)
Morphogen gradients expose cells to different signal concentrations and induce target genes with different ranges of expression. To determine how the Nodal morphogen gradient induces distinct gene expression patterns during zebrafish embryogenesis, we measured the activation dynamics of the signal
Soma Vignarajan et al.
Oncotarget, 5(15), 6289-6299 (2014-07-16)
Aberrant increase in pAKT, due to a gain-of-function mutation of PI3K or loss-of-function mutation or deletion of PTEN, occurs in prostate cancer and is associated with poor patient prognosis. Cytosolic phospholipase A₂α (cPLA₂α) is a lipid modifying enzyme by catalyzing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service